Gene Transcripts

Gene Symbol
OR4D6
Entrez Gene ID
219983
Gene Name
olfactory receptor family 4 subfamily D member 6
Gene Aliases
OR11-250
Chromosome Location
Chr. 11: 59456961 - 59457905 on Build GRCh38
UniGene ID
Hs.553650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004708.1 NP_001004708.1 1 618
GenBank mRNA BC137028.1 - 1 641
BC137029.1 - 1 641 9021025

View Full Product Details