Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301015.1 | NP_001287944.1 | 3 | 832 | |
NM_001301018.1 | NP_001287947.1 | 3 | 306 | ||
NM_001301027.1 | NP_001287956.1 | 3 | 832 | ||
NM_001301028.1 | NP_001287957.1 | 3 | 832 | ||
NM_014945.3 | NP_055760.1 | 4 | 478 | ||
XM_005268392.1 | XP_005268449.1 | 4 | 459 | ||
XM_005268393.1 | XP_005268450.1 | 4 | 459 | ||
XM_005268394.1 | XP_005268451.1 | 4 | 459 | ||
XM_005268395.1 | XP_005268452.1 | 4 | 459 | ||
XM_005268396.1 | XP_005268453.1 | 4 | 459 | ||
XM_011537609.2 | XP_011535911.1 | 4 | 459 | ||
XM_017009238.1 | XP_016864727.1 | 3 | 553 | ||
GenBank mRNA | AB020650.1 | - | 4 | 391 | |
AK225769.1 | - | 3 | 700 | ||
AK289736.1 | - | 3 | 245 | ||
BC001665.2 | - | 3 | 494 | 2988043 | |
BP285049.1 | - | 4 | 478 | ||
DQ413174.1 | - | 3 | 268 |