Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001631.4 | NP_001622.2 | 10 | 1302 | |
GenBank mRNA | AK313163.1 | - | 10 | 1291 | |
BC132678.1 | - | 10 | 1244 | 40146700 | |
M15694.1 | - | 10 | 1214 | ||
M31008.1 | - | 9 | 1304 |