Gene Transcripts

Gene Symbol
SLC39A2
Entrez Gene ID
29986
Gene Name
solute carrier family 39 member 2
Gene Aliases
6A1, ETI-1, ZIP-2, ZIP2
Chromosome Location
Chr. 14: 20999255 - 21001875 on Build GRCh38
UniGene ID
Hs.175783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256588.1 NP_001243517.1 4 861
NM_014579.3 NP_055394.2 4 866
GenBank mRNA AF186081.1 - 4 810
AK314974.1 - 4 830
AY029483.1 - 1 128
BC096723.1 - 4 697 40004409
BC098254.1 - 4 697 40004407
BC110057.1 - 4 697 40004405
D83039.1 - 4 804

View Full Product Details