Gene Transcripts

Gene Symbol
ARSI
Entrez Gene ID
340075
Gene Name
arylsulfatase family member I
Gene Aliases
SPG66
Chromosome Location
Chr. 5: 150296346 - 150302962 on Build GRCh38
UniGene ID
Hs.591252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012301.2 NP_001012301.1 2 1005
GenBank mRNA AB448735.1 - 2 996
AK122641.1 - 2 149
AY875937.1 - 2 416
BC129995.1 - 2 473 40127932
BC129996.1 - 2 473 40127934
DA264664.1 - 2 135

View Full Product Details