Gene Transcripts

Gene Symbol
IGFBP7
Entrez Gene ID
3490
Gene Name
insulin like growth factor binding protein 7
Gene Aliases
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF
Chromosome Location
Chr. 4: 57031071 - 57110385 on Build GRCh38
UniGene ID
Hs.479808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253835.1 NP_001240764.1 3 677
NM_001553.2 NP_001544.1 3 677
GenBank mRNA AK303915.1 - 3 679
AK316082.1 - 3 677
BC017201.2 - 3 665 3632247
BC066339.1 - 3 677 5296697
BT006654.1 - 3 643
L19182.1 - 3 656
S75725.1 - 3 665

View Full Product Details