Gene Transcripts

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
Chromosome Location
Chr. 5: 43486699 - 43515433 on Build GRCh38
UniGene ID
Hs.661641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198566.3 NP_940968.1 7 1620
XM_011514038.2 XP_011512340.1 7 1529
XM_017009445.1 XP_016864934.1 6 1651
XM_017009446.1 XP_016864935.1 5 1561
GenBank mRNA AK056925.1 - 7 1535
BC036867.1 - 7 1483 5198039

View Full Product Details