Gene Transcripts
- Gene Symbol
- WASH1
- Entrez Gene ID
- 100287171
- Gene Name
- WAS protein family homolog 1
- Gene Aliases
- FAM39E, WASH
- Chromosome Location
- Chr. 9: 14475 - 30487 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
- Gene Symbol
- WASH2P
- Entrez Gene ID
- 375260
- Gene Name
- WAS protein family homolog 2 pseudogene
- Gene Aliases
- FAM39B
- Chromosome Location
- Chr. 2: 113583653 - 113599037 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY217347.1 |
- |
1 |
775 |
|
BC082240.1 |
- |
1 |
267 |
|
- Gene Symbol
- WASH3P
- Entrez Gene ID
- 374666
- Gene Name
- WAS protein family homolog 3 pseudogene
- Gene Aliases
- FAM39DP
- Chromosome Location
- Chr. 15: 101960813 - 101976605 on Build GRCh38
- UniGene ID
- Hs.459573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097269.1 |
- |
1 |
945 |
|
AK300161.1 |
- |
3 |
331 |
|
BC063555.1 |
- |
1 |
250 |
|
- Gene Symbol
- WASH7P
- Entrez Gene ID
- 653635
- Gene Name
- WAS protein family homolog 7 pseudogene
- Gene Aliases
- FAM39F, WASH5P
- Chromosome Location
- Chr. 1: 14362 - 29370 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
View Full Product Details