Gene Transcripts
- Gene Symbol
- C17orf97
- Entrez Gene ID
- 400566
- Gene Name
- chromosome 17 open reading frame 97
- Gene Aliases
- CK20, LIAT1
- Chromosome Location
- Chr. 17: 410327 - 414666 on Build GRCh38
- UniGene ID
- Hs.499607
- Species
- Homo sapiens
- Gene Symbol
- LOC105371430
- Entrez Gene ID
- 105371430
- Gene Name
- uncharacterized LOC105371430
- Gene Aliases
- Chromosome Location
- Chr. 17: 404535 - 414023 on Build GRCh38
- UniGene ID
- Hs.616427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136407.1 |
|
2 |
259 |
|
View Full Product Details