Gene Transcripts
- Gene Symbol
- EVL
- Entrez Gene ID
- 51466
- Gene Name
- Enah/Vasp-like
- Gene Aliases
- RNB6
- Chromosome Location
- Chr. 14: 99971454 - 100144236 on Build GRCh38
- UniGene ID
- Hs.125867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016337.2 |
NP_057421.1
|
2, 3 |
269 |
|
XM_005267749.3 |
XP_005267806.1
|
2, 3 |
888 |
|
XM_011536828.2 |
XP_011535130.1
|
2, 3 |
902 |
|
XM_017021363.1 |
XP_016876852.1
|
2, 3 |
888 |
|
XM_017021364.1 |
XP_016876853.1
|
2, 3 |
775 |
|
GenBank mRNA |
AF052504.1 |
- |
2, 3 |
243 |
|
AF087843.1 |
- |
2, 3 |
248 |
|
AF112209.1 |
- |
2, 3 |
362 |
|
AF131766.1 |
- |
|
16 |
|
AK289720.1 |
- |
3, 4 |
279 |
|
AK295919.1 |
- |
2, 3 |
477 |
|
AK301379.1 |
- |
2, 3 |
261 |
|
BC023997.2 |
- |
2, 3 |
418 |
2822488 |
BC032358.1 |
- |
2, 3 |
240 |
5222624 |
BQ083568.1 |
- |
2, 3 |
269 |
|
View Full Product Details