Gene Transcripts

Gene Symbol
LRRC3DN
Entrez Gene ID
54083
Gene Name
LRRC3 downstream neighbor (non-protein coding)
Gene Aliases
C21orf30
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.542533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL117578.1 - 1 724

View Full Product Details