Gene Transcripts

Gene Symbol
WDR18
Entrez Gene ID
57418
Gene Name
WD repeat domain 18
Gene Aliases
Ipi3, R32184_1
Chromosome Location
Chr. 19: 982980 - 994570 on Build GRCh38
UniGene ID
Hs.325321
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024100.3 NP_077005.2 6, 7 840
XM_017027027.1 XP_016882516.1 8, 9 1366
GenBank mRNA BC000040.2 - 6, 7 825
BC001648.2 - 6, 7 820 2819452

View Full Product Details