Gene Transcripts
- Gene Symbol
- SAV1
- Entrez Gene ID
- 60485
- Gene Name
- salvador family WW domain containing protein 1
- Gene Aliases
- SAV, WW45, WWP4
- Chromosome Location
- Chr. 14: 50633106 - 50668353 on Build GRCh38
- UniGene ID
- Hs.642842
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021818.3 |
NP_068590.1
|
3, 4 |
1182 |
|
XM_011537057.2 |
XP_011535359.1
|
4, 5 |
1245 |
|
XM_017021568.1 |
XP_016877057.1
|
4, 5 |
1475 |
|
GenBank mRNA |
AJ292969.1 |
- |
3, 4 |
961 |
|
AK021500.1 |
- |
3, 4 |
1129 |
|
AK023071.1 |
- |
3, 4 |
1011 |
|
AK095903.1 |
- |
4, 5 |
1046 |
|
AK225896.1 |
- |
3, 4 |
1154 |
|
AK290883.1 |
- |
4, 5 |
1054 |
|
AL833378.1 |
- |
2, 3 |
592 |
|
BC020537.1 |
- |
3, 4 |
1128 |
4425620 |
CR457297.1 |
- |
3, 4 |
796 |
|
View Full Product Details