Gene Transcripts

Gene Symbol
RNH1
Entrez Gene ID
6050
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RAI, RNH
Chromosome Location
Chr. 11: 494512 - 507283 on Build GRCh38
UniGene ID
Hs.530687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002939.3 NP_002930.2 6 1003
NM_203383.1 NP_976317.1 6 935
NM_203384.1 NP_976318.1 5 762
NM_203385.1 NP_976319.1 5 830
NM_203386.2 NP_976320.1 6 893
NM_203387.2 NP_976321.1 6 961
NM_203388.2 NP_976322.1 5 720
NM_203389.2 NP_976323.1 5 788
XM_011520255.1 XP_011518557.1 6 938
XM_011520257.2 XP_011518559.1 6 1186
XM_011520258.2 XP_011518560.1 6 1154
XM_011520259.2 XP_011518561.1 5 703
XM_011520260.2 XP_011518562.1 6 862
XM_011520261.2 XP_011518563.1 6 936
XM_011520262.2 XP_011518564.1 6 1119
XM_011520263.1 XP_011518565.1 5 670
XM_017018106.1 XP_016873595.1 6 1003
GenBank mRNA AB209196.1 - 4 2253
AL161967.1 - 4 1938
AL534410.3 - 5 776
AL539148.3 - 5 804
BC000677.2 - 5 690 3349381
BC003506.1 - 5 703 2959514
BC011186.1 - 4 488
BC011500.1 - 5 641 4155804
BC014629.2 - 5 690 3029730
BC024037.1 - 5 703 2959514
BC047730.1 - 6 982 5788220
BM562998.1 - 5 761
BQ927358.1 - 5 649
M22414.1 - 5 651
M36717.1 - 6 890
X13973.1 - 6 890

View Full Product Details