Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 4 | 460 | |
NM_001136232.2 | NP_001129704.1 | 3 | 314 | ||
NM_001278946.1 | NP_001265875.1 | 3 | 282 | ||
NM_030673.3 | NP_109598.2 | 4 | 316 | ||
NM_183352.2 | NP_899195.1 | 3 | 282 | ||
XM_005265379.2 | XP_005265436.1 | 4 | 333 | ||
XM_017007019.1 | XP_016862508.1 | 4 | 370 | ||
XM_017007020.1 | XP_016862509.1 | 3 | 177 | ||
XM_017007021.1 | XP_016862510.1 | 3 | 187 | ||
GenBank mRNA | AB209554.1 | - | 2 | 129 | |
AF052155.1 | - | 3 | 314 | ||
AK223019.1 | - | 3 | 167 | ||
AK294662.1 | - | 3 | 187 | ||
AK301999.1 | - | 4 | 384 | ||
BC002634.2 | - | 4 | 197 | 3605545 | |
BC091506.1 | - | 3 | 192 | 30408210 | |
BE621430.1 | - | 3 | 268 | ||
CN344033.1 | - | 3 | 294 | ||
EF036494.1 | - | 3 | 181 |