Gene Transcripts

Gene Symbol
CBLN3
Entrez Gene ID
643866
Gene Name
cerebellin 3 precursor
Gene Aliases
PRO1486
Chromosome Location
Chr. 14: 24426532 - 24429525 on Build GRCh38
UniGene ID
Hs.207603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039771.2 NP_001034860.1 3 909
GenBank mRNA AY359070.1 - 3 909

View Full Product Details