Gene Transcripts

Gene Symbol
MRPL11
Entrez Gene ID
65003
Gene Name
mitochondrial ribosomal protein L11
Gene Aliases
CGI-113, L11MT, MRP-L11
Chromosome Location
Chr. 11: 66435075 - 66438860 on Build GRCh38
UniGene ID
Hs.418450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016050.4 NP_057134.1 3 341
NM_170738.3 NP_733934.1 3 263
NM_170739.3 NP_733935.1 3 341
NR_120588.1 3 333
NR_120589.1 3 191
XM_017018135.1 XP_016873624.1 3 275
GenBank mRNA AB049638.1 - 3 313
AF151871.1 - 3 325
AK127603.1 - 3 950
AK223112.1 - 3 317
AK290084.1 - 3 319
BC005002.1 - 3 320 3542758
BC108277.1 - 3 236 4416083
BI193255.1 - 3 255
BM554886.1 - 3 303
BU597133.1 - 3 344

View Full Product Details