Gene Transcripts
- Gene Symbol
- ZNF195
- Entrez Gene ID
- 7748
- Gene Name
- zinc finger protein 195
- Gene Aliases
- HRF1, ZNFP104
- Chromosome Location
- Chr. 11: 3357927 - 3379222 on Build GRCh38
- UniGene ID
- Hs.386294
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242843.1 |
NP_001229772.1
|
4, 5 |
478 |
|
NM_001256825.1 |
NP_001243754.1
|
4, 5 |
497 |
|
NM_007152.4 |
NP_009083.2
|
3, 4 |
398 |
|
NR_040083.1 |
|
5, 6 |
762 |
|
NR_046382.1 |
|
4, 5 |
584 |
|
XM_006718306.1 |
XP_006718369.1
|
4, 5 |
601 |
|
GenBank mRNA |
AK095720.1 |
- |
3, 4 |
321 |
|
AK131296.1 |
- |
5, 6 |
681 |
|
AL833722.1 |
- |
3, 4 |
311 |
|
BC121028.1 |
- |
3, 4 |
254 |
40117443 |
CX867141.1 |
- |
4, 5 |
441 |
|
DA170927.1 |
- |
4, 5 |
462 |
|
DA618142.1 |
- |
4, 5 |
414 |
|
DA984929.1 |
- |
4, 5 |
478 |
|
View Full Product Details