Gene Transcripts
- Gene Symbol
- ZFAND5
- Entrez Gene ID
- 7763
- Gene Name
- zinc finger AN1-type containing 5
- Gene Aliases
- ZA20D2, ZFAND5A, ZNF216
- Chromosome Location
- Chr. 9: 72351425 - 72365247 on Build GRCh38
- UniGene ID
- Hs.406096
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA837106.1 |
- |
2, 3 |
188 |
|
View Full Product Details