Gene Transcripts

Gene Symbol
C7orf25
Entrez Gene ID
79020
Gene Name
chromosome 7 open reading frame 25
Gene Aliases
Chromosome Location
Chr. 7: 42909273 - 42913161 on Build GRCh38
UniGene ID
Hs.698120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099858.1 NP_001093328.1 2 1224
NM_024054.2 NP_076959.2 2 1219
XM_005249844.3 XP_005249901.1 2 1494
XM_017012596.1 XP_016868085.1 2 1454
XM_017012597.1 XP_016868086.1 2 1986
GenBank mRNA AK024820.1 - 2 1192
AK225194.1 - 2 1198
AK298865.1 - 2 1224
BC000769.2 - 2 1183 2964418
BC001845.2 - 2 1183 2964418

View Full Product Details