Gene Transcripts

Gene Symbol
BBOF1
Entrez Gene ID
80127
Gene Name
basal body orientation factor 1
Gene Aliases
C14orf45, CCDC176
Chromosome Location
Chr. 14: 74019350 - 74078351 on Build GRCh38
UniGene ID
Hs.644621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025057.2 NP_079333.2 8 1380
XM_005268092.3 XP_005268149.1 8 1386
XM_005268098.4 XP_005268155.1 6 1019
XM_011537170.2 XP_011535472.1 9 1494
XM_011537171.2 XP_011535473.1 9 1494
XM_011537172.1 XP_011535474.1 9 1494
XM_011537174.2 XP_011535476.1 9 1494
XM_011537175.2 XP_011535477.1 8 1350
XM_011537176.2 XP_011535478.1 10 1478
XM_011537177.2 XP_011535479.1 8 1198
XM_011537178.2 XP_011535480.1 8 1169
XM_011537179.2 XP_011535481.1 7 1054
XM_017021659.1 XP_016877148.1 8 1386
XM_017021660.1 XP_016877149.1 7 1102
XM_017021661.1 XP_016877150.1 7 1154
XM_017021662.1 XP_016877151.1 6 938
GenBank mRNA AK026842.1 - 1 413
AK097531.1 - 6 1058
AL834487.1 - 3 655
BC104979.1 - 1 411
BC104981.1 - 1 411

View Full Product Details