Gene Transcripts

Gene Symbol
SPHKAP
Entrez Gene ID
80309
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
SKIP
Chromosome Location
Chr. 2: 227979954 - 228181999 on Build GRCh38
UniGene ID
Hs.436306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142644.1 NP_001136116.1 7 2410
NM_030623.3 NP_085126.2 7 2410
XM_005246870.4 XP_005246927.1 8 2882
XM_006712777.3 XP_006712840.1 8 2843
XM_006712778.3 XP_006712841.1 8 2842
XM_011511925.2 XP_011510227.1 9 2961
GenBank mRNA AB051465.1 - 1 1258
AK127071.1 - 7 2460
BC104954.1 - 7 2410 8143957
BC104956.1 - 7 2410 8143959
BC143408.1 - 7 2410
CR749297.1 - 1 1516
CR749494.1 - 7 2392

View Full Product Details