Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032811.2 | NP_116200.2 | 9 | 1352 | |
NR_016021.1 | 8 | 1215 | |||
XM_005271709.3 | XP_005271766.1 | 6 | 1158 | ||
XM_017018442.1 | XP_016873931.1 | 6 | 2173 | ||
GenBank mRNA | AK027527.1 | - | 8 | 1186 | |
AK074140.1 | - | 8 | 939 | ||
AK222936.1 | - | 8 | 1191 | ||
AL831881.1 | - | 3 | 192 | ||
AY696294.1 | - | 7 | 720 | ||
BC041043.1 | - | 8 | 1235 | ||
BC064392.1 | - | 8 | 1164 | ||
BC107428.1 | - | 8 | 1205 | ||
BC109269.2 | - | 7 | 834 | ||
DQ144542.1 | - | 9 | 1114 |