Gene Transcripts
- Gene Symbol
- LDHAL6B
- Entrez Gene ID
- 92483
- Gene Name
- lactate dehydrogenase A like 6B
- Gene Aliases
- LDH6B, LDHAL6, LDHL
- Chromosome Location
- Chr. 15: 59206816 - 59208588 on Build GRCh38
- UniGene ID
- Hs.307052
- Species
- Homo sapiens
- Gene Symbol
- LDHAL6FP
- Entrez Gene ID
- 100420477
- Gene Name
- lactate dehydrogenase A like 6F, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 6: 157298627 - 157300251 on Build GRCh38
- UniGene ID
- Hs.689474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127961.1 |
- |
1 |
579 |
|
View Full Product Details