Gene Transcripts

Gene Symbol
SECISBP2L
Entrez Gene ID
9728
Gene Name
SECIS binding protein 2 like
Gene Aliases
SBP2L, SLAN
Chromosome Location
Chr. 15: 48988638 - 49046563 on Build GRCh38
UniGene ID
Hs.9997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193489.1 NP_001180418.1 18 3090
NM_014701.3 NP_055516.2 17 2955
GenBank mRNA BC033001.2 - 18 3025
D87445.3 - 17 2825

View Full Product Details