Gene Transcripts

Gene Symbol
SCRN1
Entrez Gene ID
9805
Gene Name
secernin 1
Gene Aliases
SES1
Chromosome Location
Chr. 7: 29920103 - 29990289 on Build GRCh38
UniGene ID
Hs.520740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145513.1 NP_001138985.1 4 637
NM_001145514.1 NP_001138986.1 4 569
NM_001145515.1 NP_001138987.1 3 469
NM_014766.4 NP_055581.3 4 629
GenBank mRNA AB071705.1 - 4 519
AK289514.1 - 3 664
AK295172.1 - 4 569
AK295713.1 - 3 372
BC040492.1 - 4 581 5314711
D83777.2 - 4 574

View Full Product Details