Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67663425-67663917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B2
Entrez Gene ID
222
Gene Name
aldehyde dehydrogenase 3 family member B2
Gene Aliases
ALDH8
UniGene
Hs.87539
Gene Chromosome Location
Chr.11: 67662162-67681214 on Build GRCh38
Transcripts
NM_000695.3 NM_001031615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2042329 c.933C>T p.F311F Chr.11: 67663702-67663702 on Build GRCh38
6224828 c.918A>G p.G306G Chr.11: 67663717-67663717 on Build GRCh38
6885071 c.953C>T p.S318F Chr.11: 67663682-67663682 on Build GRCh38
7319043 c.963C>T p.F321F Chr.11: 67663672-67663672 on Build GRCh38
6482705 c.895C>A p.R299R Chr.11: 67663740-67663740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761283003 Chr.11: 67663717-67663717 on Build GRCh38
rs764590976 Chr.11: 67663623-67663623 on Build GRCh38
rs754902808 Chr.11: 67663672-67663672 on Build GRCh38
rs749667333 Chr.11: 67663701-67663701 on Build GRCh38
rs767153238 Chr.11: 67663647-67663647 on Build GRCh38
rs200921334 Chr.11: 67663764-67663764 on Build GRCh38
rs375224798 Chr.11: 67663681-67663681 on Build GRCh38
rs1037704224 Chr.11: 67663596-67663596 on Build GRCh38
rs779431883 Chr.11: 67663470-67663470 on Build GRCh38
rs940181779 Chr.11: 67663592-67663592 on Build GRCh38
rs753669020 Chr.11: 67663732-67663732 on Build GRCh38
rs769140741 Chr.11: 67663713-67663713 on Build GRCh38
rs780382602 Chr.11: 67663769-67663769 on Build GRCh38
rs769663166 Chr.11: 67663836-67663836 on Build GRCh38
rs535400854 Chr.11: 67663758-67663758 on Build GRCh38
rs1000167407 Chr.11: 67663841-67663841 on Build GRCh38
rs149164164 Chr.11: 67663696-67663696 on Build GRCh38
rs957902205 Chr.11: 67663848-67663848 on Build GRCh38
rs763244482 Chr.11: 67663461-67663461 on Build GRCh38
rs774905070 Chr.11: 67663714-67663714 on Build GRCh38
rs970657840 Chr.11: 67663862-67663862 on Build GRCh38
rs750091803 Chr.11: 67663658-67663658 on Build GRCh38
rs751014793 Chr.11: 67663632-67663632 on Build GRCh38
rs1037186700 Chr.11: 67663785-67663785 on Build GRCh38
rs774719083 Chr.11: 67663791-67663791 on Build GRCh38
rs766887079 Chr.11: 67663723-67663723 on Build GRCh38
rs749648863 Chr.11: 67663770-67663770 on Build GRCh38
rs370310827 Chr.11: 67663809-67663809 on Build GRCh38
rs1002392879 Chr.11: 67663471-67663471 on Build GRCh38
rs769049154 Chr.11: 67663788-67663788 on Build GRCh38
rs1049964198 Chr.11: 67663821-67663821 on Build GRCh38
rs752372318 Chr.11: 67663688-67663688 on Build GRCh38
rs573086275 Chr.11: 67663796-67663796 on Build GRCh38
rs899500913 Chr.11: 67663812-67663812 on Build GRCh38
rs559217458 Chr.11: 67663449-67663449 on Build GRCh38
rs775654360 Chr.11: 67663697-67663697 on Build GRCh38
rs4646826 Chr.11: 67663729-67663729 on Build GRCh38
rs748391891 Chr.11: 67663522-67663522 on Build GRCh38
rs147827325 Chr.11: 67663746-67663746 on Build GRCh38
rs141294556 Chr.11: 67663853-67663853 on Build GRCh38
rs201544486 Chr.11: 67663798-67663798 on Build GRCh38
rs765018419 Chr.11: 67663641-67663641 on Build GRCh38
rs748614000 Chr.11: 67663795-67663795 on Build GRCh38
rs996647969 Chr.11: 67663839-67663839 on Build GRCh38
rs544480597 Chr.11: 67663877-67663877 on Build GRCh38
rs929425243 Chr.11: 67663772-67663772 on Build GRCh38
rs781457219 Chr.11: 67663691-67663691 on Build GRCh38
rs368813208 Chr.11: 67663624-67663624 on Build GRCh38
rs141299989 Chr.11: 67663750-67663750 on Build GRCh38
rs1020900320 Chr.11: 67663856-67663856 on Build GRCh38
rs200176520 Chr.11: 67663671-67663671 on Build GRCh38
rs552303161 Chr.11: 67663582-67663582 on Build GRCh38
rs867378007 Chr.11: 67663649-67663649 on Build GRCh38
rs901414022 Chr.11: 67663603-67663603 on Build GRCh38
rs766047073 Chr.11: 67663663-67663663 on Build GRCh38
rs550915491 Chr.11: 67663890-67663890 on Build GRCh38
rs779048817 Chr.11: 67663680-67663680 on Build GRCh38
rs775266509 Chr.11: 67663740-67663740 on Build GRCh38
rs1033068976 Chr.11: 67663640-67663640 on Build GRCh38
rs202204174 Chr.11: 67663694-67663694 on Build GRCh38
rs770256337 Chr.11: 67663765-67663765 on Build GRCh38
rs889754911 Chr.11: 67663618-67663618 on Build GRCh38
rs61757682 Chr.11: 67663748-67663748 on Build GRCh38
rs1010524187 Chr.11: 67663889-67663889 on Build GRCh38
rs368400737 Chr.11: 67663739-67663739 on Build GRCh38
rs145285372 Chr.11: 67663662-67663662 on Build GRCh38
rs747773263 Chr.11: 67663730-67663730 on Build GRCh38
rs998212659 Chr.11: 67663604-67663604 on Build GRCh38
rs949783146 Chr.11: 67663466-67663466 on Build GRCh38
rs1979376 Chr.11: 67663639-67663639 on Build GRCh38
rs1979375 Chr.11: 67663567-67663567 on Build GRCh38
rs930387891 Chr.11: 67663562-67663562 on Build GRCh38
rs180859869 Chr.11: 67663728-67663728 on Build GRCh38
rs889808829 Chr.11: 67663835-67663835 on Build GRCh38
rs758040222 Chr.11: 67663712-67663712 on Build GRCh38
rs113365834 Chr.11: 67663615-67663615 on Build GRCh38
rs1033538605 Chr.11: 67663842-67663842 on Build GRCh38
rs777333106 Chr.11: 67663783-67663784 on Build GRCh38
rs775948429 Chr.11: 67663451-67663451 on Build GRCh38

View Full Product Details