Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67666486-67666992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B2
Entrez Gene ID
222
Gene Name
aldehyde dehydrogenase 3 family member B2
Gene Aliases
ALDH8
UniGene
Hs.87539
Gene Chromosome Location
Chr.11: 67662162-67681214 on Build GRCh38
Transcripts
NM_000695.3 NM_001031615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3452376 c.2T>G p.M1R Chr.11: 67666934-67666934 on Build GRCh38
255677 c.48G>T p.S16S Chr.11: 67666677-67666677 on Build GRCh38
6598278 c.92C>T p.A31V Chr.11: 67666633-67666633 on Build GRCh38
4599442 c.148G>A p.A50T Chr.11: 67666577-67666577 on Build GRCh38
5584425 c.90C>T p.I30I Chr.11: 67666635-67666635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914896990 Chr.11: 67666542-67666542 on Build GRCh38
rs878866410 Chr.11: 67666641-67666641 on Build GRCh38
rs112877473 Chr.11: 67666692-67666692 on Build GRCh38
rs777455441 Chr.11: 67666941-67666941 on Build GRCh38
rs749766899 Chr.11: 67666724-67666724 on Build GRCh38
rs781026325 Chr.11: 67666951-67666951 on Build GRCh38
rs772103224 Chr.11: 67666962-67666962 on Build GRCh38
rs189634559 Chr.11: 67666920-67666920 on Build GRCh38
rs1007374366 Chr.11: 67666883-67666883 on Build GRCh38
rs745451949 Chr.11: 67666699-67666699 on Build GRCh38
rs767796810 Chr.11: 67666706-67666706 on Build GRCh38
rs757928272 Chr.11: 67666936-67666936 on Build GRCh38
rs927758835 Chr.11: 67666678-67666678 on Build GRCh38
rs762536819 Chr.11: 67666534-67666534 on Build GRCh38
rs774970209 Chr.11: 67666567-67666567 on Build GRCh38
rs1051493 Chr.11: 67666616-67666616 on Build GRCh38
rs773768354 Chr.11: 67666649-67666649 on Build GRCh38
rs376886394 Chr.11: 67666742-67666742 on Build GRCh38
rs755502152 Chr.11: 67666914-67666914 on Build GRCh38
rs765146530 Chr.11: 67666880-67666880 on Build GRCh38
rs758550912 Chr.11: 67666598-67666598 on Build GRCh38
rs759422757 Chr.11: 67666685-67666685 on Build GRCh38
rs762591374 Chr.11: 67666569-67666569 on Build GRCh38
rs758540346 Chr.11: 67666554-67666554 on Build GRCh38
rs201115308 Chr.11: 67666555-67666555 on Build GRCh38
rs968620332 Chr.11: 67666910-67666910 on Build GRCh38
rs1140671 Chr.11: 67666580-67666580 on Build GRCh38
rs76550666 Chr.11: 67666690-67666690 on Build GRCh38
rs1038382731 Chr.11: 67666796-67666796 on Build GRCh38
rs557052250 Chr.11: 67666516-67666516 on Build GRCh38
rs184185910 Chr.11: 67666537-67666537 on Build GRCh38
rs750598695 Chr.11: 67666892-67666892 on Build GRCh38
rs372646847 Chr.11: 67666610-67666610 on Build GRCh38
rs750340117 Chr.11: 67666540-67666540 on Build GRCh38
rs1026310648 Chr.11: 67666822-67666822 on Build GRCh38
rs771278201 Chr.11: 67666558-67666558 on Build GRCh38
rs770300354 Chr.11: 67666565-67666565 on Build GRCh38
rs572991645 Chr.11: 67666912-67666912 on Build GRCh38
rs987340752 Chr.11: 67666606-67666606 on Build GRCh38
rs919325742 Chr.11: 67666512-67666512 on Build GRCh38
rs774105344 Chr.11: 67666578-67666578 on Build GRCh38
rs754196722 Chr.11: 67666970-67666970 on Build GRCh38
rs756364223 Chr.11: 67666893-67666893 on Build GRCh38
rs780579312 Chr.11: 67666621-67666621 on Build GRCh38
rs745718592 Chr.11: 67666958-67666958 on Build GRCh38
rs113398365 Chr.11: 67666918-67666918 on Build GRCh38
rs140908160 Chr.11: 67666680-67666680 on Build GRCh38
rs74508424 Chr.11: 67666607-67666607 on Build GRCh38
rs777667881 Chr.11: 67666585-67666585 on Build GRCh38
rs752789383 Chr.11: 67666596-67666596 on Build GRCh38
rs140248913 Chr.11: 67666618-67666618 on Build GRCh38
rs112071378 Chr.11: 67666602-67666602 on Build GRCh38
rs545124772 Chr.11: 67666515-67666515 on Build GRCh38
rs927109096 Chr.11: 67666769-67666769 on Build GRCh38
rs368925198 Chr.11: 67666645-67666645 on Build GRCh38
rs74946629 Chr.11: 67666562-67666562 on Build GRCh38
rs757458910 Chr.11: 67666691-67666691 on Build GRCh38
rs766488386 Chr.11: 67666587-67666587 on Build GRCh38
rs201106421 Chr.11: 67666879-67666879 on Build GRCh38
rs1038210652 Chr.11: 67666841-67666841 on Build GRCh38
rs200387182 Chr.11: 67666959-67666959 on Build GRCh38
rs755447214 Chr.11: 67666731-67666731 on Build GRCh38
rs116571818 Chr.11: 67666716-67666716 on Build GRCh38
rs544174705 Chr.11: 67666948-67666948 on Build GRCh38
rs746679144 Chr.11: 67666863-67666863 on Build GRCh38
rs757423369 Chr.11: 67666609-67666609 on Build GRCh38
rs776292851 Chr.11: 67666875-67666875 on Build GRCh38
rs1021825718 Chr.11: 67666780-67666780 on Build GRCh38
rs3741177 Chr.11: 67666553-67666553 on Build GRCh38
rs139106840 Chr.11: 67666650-67666650 on Build GRCh38
rs761711204 Chr.11: 67666538-67666538 on Build GRCh38
rs894609911 Chr.11: 67666563-67666563 on Build GRCh38
rs770808847 Chr.11: 67666871-67666871 on Build GRCh38
rs372661590 Chr.11: 67666557-67666557 on Build GRCh38
rs756154514 Chr.11: 67666543-67666543 on Build GRCh38
rs748729064 Chr.11: 67666634-67666634 on Build GRCh38
rs766393325 Chr.11: 67666658-67666658 on Build GRCh38
rs3741178 Chr.11: 67666577-67666577 on Build GRCh38
rs374258648 Chr.11: 67666886-67666886 on Build GRCh38
rs916978196 Chr.11: 67666707-67666707 on Build GRCh38
rs899401975 Chr.11: 67666800-67666800 on Build GRCh38
rs538601956 Chr.11: 67666712-67666712 on Build GRCh38
rs766571106 Chr.11: 67666969-67666969 on Build GRCh38
rs760583105 Chr.11: 67666657-67666657 on Build GRCh38
rs74761791 Chr.11: 67666677-67666677 on Build GRCh38
rs746291984 Chr.11: 67666611-67666611 on Build GRCh38
rs1051633266 Chr.11: 67666869-67666869 on Build GRCh38
rs386754500 Chr.11: 67666689-67666690 on Build GRCh38
rs779075204 Chr.11: 67666549-67666549 on Build GRCh38
rs778432741 Chr.11: 67666737-67666737 on Build GRCh38
rs141932310 Chr.11: 67666919-67666919 on Build GRCh38
rs775339680 Chr.11: 67666960-67666960 on Build GRCh38
rs1017670350 Chr.11: 67666877-67666877 on Build GRCh38
rs368232009 Chr.11: 67666630-67666630 on Build GRCh38
rs993939387 Chr.11: 67666821-67666821 on Build GRCh38
rs184650566 Chr.11: 67666722-67666722 on Build GRCh38
rs766688290 Chr.11: 67666963-67666963 on Build GRCh38
rs767382764 Chr.11: 67666581-67666581 on Build GRCh38
rs941319264 Chr.11: 67666832-67666832 on Build GRCh38
rs575524604 Chr.11: 67666519-67666519 on Build GRCh38
rs1140670 Chr.11: 67666586-67666586 on Build GRCh38
rs964503981 Chr.11: 67666612-67666612 on Build GRCh38
rs371195464 Chr.11: 67666862-67666862 on Build GRCh38
rs774986866 Chr.11: 67666525-67666525 on Build GRCh38
rs761921125 Chr.11: 67666887-67666887 on Build GRCh38
rs978376568 Chr.11: 67666896-67666896 on Build GRCh38
rs925597187 Chr.11: 67666559-67666559 on Build GRCh38
rs1871032 Chr.11: 67666539-67666539 on Build GRCh38
rs76408306 Chr.11: 67666689-67666689 on Build GRCh38
rs4646823 Chr.11: 67666945-67666945 on Build GRCh38
rs368134300 Chr.11: 67666903-67666903 on Build GRCh38

View Full Product Details