Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69703166-69703657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402940 c.284T>A p.V95E Chr.11: 69703313-69703313 on Build GRCh38
5839451 c.244A>G p.I82V Chr.11: 69703353-69703353 on Build GRCh38
1207067 c.295C>T p.R99W Chr.11: 69703302-69703302 on Build GRCh38
5477786 c.335T>C p.L112P Chr.11: 69703262-69703262 on Build GRCh38
6552349 c.220C>A p.Q74K Chr.11: 69703657-69703657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903882310 Chr.11: 69703198-69703198 on Build GRCh38
rs115437861 Chr.11: 69703224-69703224 on Build GRCh38
rs775508698 Chr.11: 69703382-69703382 on Build GRCh38
rs1013693090 Chr.11: 69703585-69703585 on Build GRCh38
rs978162555 Chr.11: 69703401-69703401 on Build GRCh38
rs555015996 Chr.11: 69703598-69703598 on Build GRCh38
rs957011175 Chr.11: 69703219-69703219 on Build GRCh38
rs867463501 Chr.11: 69703512-69703512 on Build GRCh38
rs377560349 Chr.11: 69703242-69703242 on Build GRCh38
rs774757875 Chr.11: 69703391-69703391 on Build GRCh38
rs548598569 Chr.11: 69703220-69703220 on Build GRCh38
rs768058336 Chr.11: 69703286-69703286 on Build GRCh38
rs573160787 Chr.11: 69703600-69703600 on Build GRCh38
rs989784991 Chr.11: 69703470-69703470 on Build GRCh38
rs753563763 Chr.11: 69703309-69703309 on Build GRCh38
rs1024792578 Chr.11: 69703589-69703589 on Build GRCh38
rs768545527 Chr.11: 69703214-69703214 on Build GRCh38
rs755614421 Chr.11: 69703305-69703305 on Build GRCh38
rs1046522275 Chr.11: 69703555-69703555 on Build GRCh38
rs1002113100 Chr.11: 69703607-69703607 on Build GRCh38
rs540968622 Chr.11: 69703606-69703606 on Build GRCh38
rs199757429 Chr.11: 69703221-69703221 on Build GRCh38
rs868553800 Chr.11: 69703586-69703586 on Build GRCh38
rs575836131 Chr.11: 69703508-69703508 on Build GRCh38
rs747918000 Chr.11: 69703344-69703344 on Build GRCh38
rs923596823 Chr.11: 69703429-69703429 on Build GRCh38
rs939925445 Chr.11: 69703599-69703599 on Build GRCh38
rs1001352049 Chr.11: 69703207-69703207 on Build GRCh38
rs957932691 Chr.11: 69703462-69703462 on Build GRCh38
rs948250492 Chr.11: 69703539-69703539 on Build GRCh38
rs111621405 Chr.11: 69703422-69703422 on Build GRCh38
rs769885512 Chr.11: 69703376-69703376 on Build GRCh38
rs151043816 Chr.11: 69703272-69703272 on Build GRCh38
rs969236533 Chr.11: 69703596-69703596 on Build GRCh38
rs370913329 Chr.11: 69703384-69703384 on Build GRCh38
rs777761575 Chr.11: 69703371-69703371 on Build GRCh38
rs768811599 Chr.11: 69703387-69703387 on Build GRCh38
rs769977243 Chr.11: 69703251-69703251 on Build GRCh38
rs919766948 Chr.11: 69703613-69703613 on Build GRCh38
rs972938511 Chr.11: 69703604-69703604 on Build GRCh38
rs779955147 Chr.11: 69703317-69703317 on Build GRCh38
rs771977220 Chr.11: 69703235-69703235 on Build GRCh38
rs1039969908 Chr.11: 69703502-69703502 on Build GRCh38
rs889553161 Chr.11: 69703628-69703628 on Build GRCh38
rs1049476287 Chr.11: 69703622-69703622 on Build GRCh38
rs995993538 Chr.11: 69703335-69703335 on Build GRCh38
rs112832084 Chr.11: 69703339-69703339 on Build GRCh38
rs758325107 Chr.11: 69703364-69703364 on Build GRCh38
rs892520788 Chr.11: 69703561-69703561 on Build GRCh38
rs931172575 Chr.11: 69703618-69703618 on Build GRCh38
rs779898424 Chr.11: 69703306-69703306 on Build GRCh38
rs1016220726 Chr.11: 69703633-69703633 on Build GRCh38
rs543125202 Chr.11: 69703559-69703559 on Build GRCh38
rs571752517 Chr.11: 69703246-69703246 on Build GRCh38
rs775667808 Chr.11: 69703254-69703254 on Build GRCh38
rs749971879 Chr.11: 69703287-69703287 on Build GRCh38
rs762243036 Chr.11: 69703284-69703284 on Build GRCh38
rs754635108 Chr.11: 69703312-69703312 on Build GRCh38
rs963626963 Chr.11: 69703634-69703634 on Build GRCh38
rs763115002 Chr.11: 69703257-69703257 on Build GRCh38
rs964578879 Chr.11: 69703324-69703324 on Build GRCh38
rs745984073 Chr.11: 69703374-69703374 on Build GRCh38
rs751688902 Chr.11: 69703516-69703516 on Build GRCh38
rs778799135 Chr.11: 69703216-69703216 on Build GRCh38
rs764476339 Chr.11: 69703260-69703260 on Build GRCh38

View Full Product Details