Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69704083-69704611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

SNP Detail

SNP ID Chromosome Location
rs982366457 Chr.11: 69704214-69704214 on Build GRCh38
rs920643659 Chr.11: 69704425-69704425 on Build GRCh38
rs563158441 Chr.11: 69704412-69704412 on Build GRCh38
rs893561008 Chr.11: 69704430-69704430 on Build GRCh38
rs988405881 Chr.11: 69704173-69704173 on Build GRCh38
rs1052673477 Chr.11: 69704413-69704413 on Build GRCh38
rs935190042 Chr.11: 69704411-69704411 on Build GRCh38
rs890345163 Chr.11: 69704265-69704266 on Build GRCh38
rs1010007364 Chr.11: 69704558-69704558 on Build GRCh38
rs999746180 Chr.11: 69704144-69704144 on Build GRCh38
rs914116927 Chr.11: 69704175-69704176 on Build GRCh38
rs982390294 Chr.11: 69704410-69704410 on Build GRCh38
rs951602386 Chr.11: 69704254-69704254 on Build GRCh38
rs933976560 Chr.11: 69704367-69704367 on Build GRCh38
rs1038960881 Chr.11: 69704487-69704487 on Build GRCh38
rs911751794 Chr.11: 69704456-69704456 on Build GRCh38
rs1010548268 Chr.11: 69704182-69704182 on Build GRCh38
rs955817673 Chr.11: 69704362-69704362 on Build GRCh38
rs969622553 Chr.11: 69704204-69704204 on Build GRCh38
rs926634610 Chr.11: 69704183-69704183 on Build GRCh38
rs1023219863 Chr.11: 69704191-69704191 on Build GRCh38
rs938096940 Chr.11: 69704202-69704202 on Build GRCh38
rs4075423 Chr.11: 69704530-69704530 on Build GRCh38
rs1047564836 Chr.11: 69704241-69704241 on Build GRCh38
rs978940656 Chr.11: 69704124-69704124 on Build GRCh38
rs35775831 Chr.11: 69704470-69704471 on Build GRCh38
rs115533464 Chr.11: 69704444-69704444 on Build GRCh38
rs188042408 Chr.11: 69704308-69704308 on Build GRCh38
rs958479648 Chr.11: 69704162-69704163 on Build GRCh38
rs1010114826 Chr.11: 69704363-69704363 on Build GRCh38
rs983342517 Chr.11: 69704256-69704256 on Build GRCh38
rs1030777052 Chr.11: 69704176-69704176 on Build GRCh38
rs1051703669 Chr.11: 69704379-69704379 on Build GRCh38
rs890354068 Chr.11: 69704392-69704392 on Build GRCh38
rs1008790224 Chr.11: 69704271-69704271 on Build GRCh38
rs528169884 Chr.11: 69704585-69704585 on Build GRCh38
rs542318249 Chr.11: 69704469-69704469 on Build GRCh38
rs369000431 Chr.11: 69704381-69704381 on Build GRCh38
rs552803971 Chr.11: 69704586-69704586 on Build GRCh38
rs1035232907 Chr.11: 69704234-69704234 on Build GRCh38
rs1023334937 Chr.11: 69704514-69704514 on Build GRCh38
rs964407806 Chr.11: 69704115-69704115 on Build GRCh38
rs909982187 Chr.11: 69704291-69704291 on Build GRCh38
rs975418348 Chr.11: 69704323-69704323 on Build GRCh38
rs893516556 Chr.11: 69704179-69704179 on Build GRCh38
rs1054135052 Chr.11: 69704539-69704539 on Build GRCh38
rs540664671 Chr.11: 69704162-69704162 on Build GRCh38
rs544889942 Chr.11: 69704360-69704360 on Build GRCh38
rs891510162 Chr.11: 69704551-69704551 on Build GRCh38
rs999938627 Chr.11: 69704327-69704327 on Build GRCh38
rs1021114841 Chr.11: 69704559-69704559 on Build GRCh38
rs1032497136 Chr.11: 69704127-69704127 on Build GRCh38
rs558678896 Chr.11: 69704232-69704232 on Build GRCh38
rs180906049 Chr.11: 69704568-69704568 on Build GRCh38

View Full Product Details