Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72842531-72843026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L2
Entrez Gene ID
89849
Gene Name
autophagy related 16 like 2
Gene Aliases
ATG16B, WDR80
UniGene
Hs.653186
Gene Chromosome Location
Chr.11: 72814406-72843740 on Build GRCh38
Transcripts
XM_005274376.4 NM_014824.2 XM_011545409.1 XM_011545410.2 XM_017018632.1

Gene Symbol
FCHSD2
Entrez Gene ID
9873
Gene Name
FCH and double SH3 domains 2
Gene Aliases
NWK, SH3MD3
UniGene
Hs.744959
Gene Chromosome Location
Chr.11: 72836743-73142682 on Build GRCh38
Transcripts
XM_005274376.4 NM_014824.2 XM_011545409.1 XM_011545410.2 XM_017018632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710430 c.1753G>A p.E585K Chr.11: 72842794-72842794 on Build GRCh38
294017 c.1617T>C p.R539R Chr.11: 72842762-72842762 on Build GRCh38
7182950 c.1579G>A p.D527N Chr.11: 72842800-72842800 on Build GRCh38
931765 c.1737G>T p.E579D Chr.11: 72842810-72842810 on Build GRCh38
7182951 c.1747G>A p.D583N Chr.11: 72842800-72842800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758595614 Chr.11: 72842713-72842713 on Build GRCh38
rs746187247 Chr.11: 72842625-72842625 on Build GRCh38
rs776986374 Chr.11: 72842615-72842615 on Build GRCh38
rs781752775 Chr.11: 72842760-72842760 on Build GRCh38
rs768396524 Chr.11: 72842605-72842605 on Build GRCh38
rs766244653 Chr.11: 72842840-72842840 on Build GRCh38
rs996747754 Chr.11: 72842956-72842956 on Build GRCh38
rs775282196 Chr.11: 72842639-72842639 on Build GRCh38
rs748017056 Chr.11: 72842763-72842763 on Build GRCh38
rs990779969 Chr.11: 72842739-72842739 on Build GRCh38
rs765010766 Chr.11: 72842919-72842919 on Build GRCh38
rs949487223 Chr.11: 72842752-72842752 on Build GRCh38
rs774000414 Chr.11: 72842811-72842811 on Build GRCh38
rs748966822 Chr.11: 72842792-72842792 on Build GRCh38
rs770238136 Chr.11: 72842764-72842764 on Build GRCh38
rs768378654 Chr.11: 72842795-72842795 on Build GRCh38
rs192398642 Chr.11: 72842881-72842881 on Build GRCh38
rs144200516 Chr.11: 72842680-72842680 on Build GRCh38
rs755994711 Chr.11: 72842704-72842704 on Build GRCh38
rs751572265 Chr.11: 72842718-72842718 on Build GRCh38
rs760497980 Chr.11: 72842838-72842838 on Build GRCh38
rs373737720 Chr.11: 72842634-72842634 on Build GRCh38
rs757397648 Chr.11: 72842885-72842885 on Build GRCh38
rs771693765 Chr.11: 72842833-72842833 on Build GRCh38
rs532041123 Chr.11: 72842583-72842583 on Build GRCh38
rs555714802 Chr.11: 72843003-72843003 on Build GRCh38
rs570031472 Chr.11: 72842629-72842629 on Build GRCh38
rs747538666 Chr.11: 72842609-72842609 on Build GRCh38
rs576622316 Chr.11: 72842886-72842886 on Build GRCh38
rs147782509 Chr.11: 72842703-72842703 on Build GRCh38
rs190366587 Chr.11: 72842969-72842969 on Build GRCh38
rs751768510 Chr.11: 72842853-72842853 on Build GRCh38
rs929120074 Chr.11: 72842855-72842855 on Build GRCh38
rs895684977 Chr.11: 72842767-72842767 on Build GRCh38
rs1018025742 Chr.11: 72842926-72842926 on Build GRCh38
rs200377316 Chr.11: 72842777-72842777 on Build GRCh38
rs201613221 Chr.11: 72842846-72842846 on Build GRCh38
rs900704464 Chr.11: 72842950-72842950 on Build GRCh38
rs763166449 Chr.11: 72842847-72842847 on Build GRCh38
rs370065778 Chr.11: 72842612-72842612 on Build GRCh38
rs758540990 Chr.11: 72842603-72842603 on Build GRCh38
rs766081890 Chr.11: 72842709-72842709 on Build GRCh38
rs764245364 Chr.11: 72842850-72842850 on Build GRCh38
rs867398749 Chr.11: 72842997-72842997 on Build GRCh38
rs761346030 Chr.11: 72842831-72842831 on Build GRCh38
rs761869339 Chr.11: 72842687-72842687 on Build GRCh38
rs527297703 Chr.11: 72842848-72842848 on Build GRCh38
rs767390327 Chr.11: 72842575-72842575 on Build GRCh38
rs927172693 Chr.11: 72842666-72842666 on Build GRCh38
rs913325125 Chr.11: 72842570-72842570 on Build GRCh38
rs1005331492 Chr.11: 72842906-72842906 on Build GRCh38
rs777862692 Chr.11: 72842717-72842717 on Build GRCh38
rs762486252 Chr.11: 72842665-72842665 on Build GRCh38
rs200391084 Chr.11: 72842750-72842750 on Build GRCh38
rs773945981 Chr.11: 72842685-72842685 on Build GRCh38
rs371718185 Chr.11: 72842789-72842789 on Build GRCh38
rs750992653 Chr.11: 72842887-72842887 on Build GRCh38
rs748303731 Chr.11: 72842584-72842584 on Build GRCh38

View Full Product Details