Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77245203-77245705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD4
Entrez Gene ID
220032
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases
GDE6
UniGene
Hs.249795
Gene Chromosome Location
Chr.11: 77216558-77301699 on Build GRCh38
Transcripts
NM_182833.1 XM_011544826.1 XM_011544833.1 XM_011544834.1 XM_017017348.1 XM_017017349.1 XM_017017350.1 XM_017017351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6215348 c.979T>C p.F327L Chr.11: 77245388-77245388 on Build GRCh38
932163 c.977A>C p.K326T Chr.11: 77245390-77245390 on Build GRCh38
4686752 c.1040G>A p.R347H Chr.11: 77245327-77245327 on Build GRCh38
5612471 c.1019C>T p.P340L Chr.11: 77245348-77245348 on Build GRCh38
3453499 c.1018C>T p.P340S Chr.11: 77245349-77245349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755647838 Chr.11: 77245552-77245552 on Build GRCh38
rs759809373 Chr.11: 77245373-77245373 on Build GRCh38
rs151093063 Chr.11: 77245555-77245555 on Build GRCh38
rs143740353 Chr.11: 77245422-77245422 on Build GRCh38
rs780407258 Chr.11: 77245314-77245314 on Build GRCh38
rs747412510 Chr.11: 77245450-77245450 on Build GRCh38
rs371285012 Chr.11: 77245369-77245369 on Build GRCh38
rs748630571 Chr.11: 77245621-77245621 on Build GRCh38
rs111333251 Chr.11: 77245578-77245579 on Build GRCh38
rs773785529 Chr.11: 77245448-77245448 on Build GRCh38
rs557032963 Chr.11: 77245514-77245514 on Build GRCh38
rs866365678 Chr.11: 77245268-77245268 on Build GRCh38
rs762247508 Chr.11: 77245337-77245337 on Build GRCh38
rs372911205 Chr.11: 77245556-77245556 on Build GRCh38
rs766316923 Chr.11: 77245367-77245367 on Build GRCh38
rs766657500 Chr.11: 77245293-77245293 on Build GRCh38
rs748557699 Chr.11: 77245432-77245432 on Build GRCh38
rs552942099 Chr.11: 77245328-77245328 on Build GRCh38
rs539232786 Chr.11: 77245562-77245562 on Build GRCh38
rs777161880 Chr.11: 77245608-77245608 on Build GRCh38
rs757199465 Chr.11: 77245531-77245531 on Build GRCh38
rs751365584 Chr.11: 77245516-77245516 on Build GRCh38
rs774914693 Chr.11: 77245335-77245335 on Build GRCh38
rs1030164666 Chr.11: 77245587-77245587 on Build GRCh38
rs753904021 Chr.11: 77245610-77245610 on Build GRCh38
rs753961791 Chr.11: 77245302-77245302 on Build GRCh38
rs771451998 Chr.11: 77245453-77245453 on Build GRCh38
rs758148147 Chr.11: 77245396-77245396 on Build GRCh38
rs778742764 Chr.11: 77245309-77245309 on Build GRCh38
rs765419939 Chr.11: 77245382-77245382 on Build GRCh38
rs937675089 Chr.11: 77245425-77245425 on Build GRCh38
rs553582346 Chr.11: 77245545-77245545 on Build GRCh38
rs1053492041 Chr.11: 77245342-77245342 on Build GRCh38
rs980976500 Chr.11: 77245631-77245631 on Build GRCh38
rs560775485 Chr.11: 77245228-77245228 on Build GRCh38
rs760643120 Chr.11: 77245271-77245271 on Build GRCh38
rs745372030 Chr.11: 77245421-77245421 on Build GRCh38
rs151233592 Chr.11: 77245327-77245327 on Build GRCh38
rs757755613 Chr.11: 77245612-77245612 on Build GRCh38
rs772519926 Chr.11: 77245433-77245433 on Build GRCh38
rs750581350 Chr.11: 77245267-77245267 on Build GRCh38
rs773555890 Chr.11: 77245352-77245352 on Build GRCh38
rs575951657 Chr.11: 77245673-77245673 on Build GRCh38
rs936397952 Chr.11: 77245669-77245669 on Build GRCh38
rs772747233 Chr.11: 77245348-77245348 on Build GRCh38
rs777758542 Chr.11: 77245268-77245269 on Build GRCh38
rs878870688 Chr.11: 77245460-77245460 on Build GRCh38
rs962688108 Chr.11: 77245654-77245654 on Build GRCh38
rs376353628 Chr.11: 77245426-77245426 on Build GRCh38
rs998006146 Chr.11: 77245583-77245583 on Build GRCh38
rs749358780 Chr.11: 77245325-77245325 on Build GRCh38
rs752527111 Chr.11: 77245394-77245394 on Build GRCh38
rs771015641 Chr.11: 77245335-77245336 on Build GRCh38
rs926354359 Chr.11: 77245641-77245641 on Build GRCh38
rs749675511 Chr.11: 77245551-77245553 on Build GRCh38
rs140271685 Chr.11: 77245366-77245366 on Build GRCh38
rs772435235 Chr.11: 77245482-77245483 on Build GRCh38
rs77289256 Chr.11: 77245357-77245357 on Build GRCh38
rs7938075 Chr.11: 77245604-77245604 on Build GRCh38
rs951628636 Chr.11: 77245229-77245229 on Build GRCh38
rs532591376 Chr.11: 77245313-77245313 on Build GRCh38
rs746559076 Chr.11: 77245319-77245319 on Build GRCh38
rs567924678 Chr.11: 77245488-77245488 on Build GRCh38
rs775004469 Chr.11: 77245245-77245245 on Build GRCh38
rs759578408 Chr.11: 77245473-77245473 on Build GRCh38
rs779125944 Chr.11: 77245360-77245361 on Build GRCh38
rs369664698 Chr.11: 77245461-77245461 on Build GRCh38
rs775715309 Chr.11: 77245361-77245361 on Build GRCh38
rs983442275 Chr.11: 77245230-77245230 on Build GRCh38
rs764040453 Chr.11: 77245397-77245397 on Build GRCh38
rs766292874 Chr.11: 77245359-77245359 on Build GRCh38
rs747692990 Chr.11: 77245452-77245455 on Build GRCh38
rs368753721 Chr.11: 77245304-77245304 on Build GRCh38
rs779877916 Chr.11: 77245553-77245553 on Build GRCh38
rs374487421 Chr.11: 77245237-77245237 on Build GRCh38
rs995904742 Chr.11: 77245541-77245541 on Build GRCh38
rs751394717 Chr.11: 77245409-77245409 on Build GRCh38
rs775639161 Chr.11: 77245490-77245490 on Build GRCh38
rs762748425 Chr.11: 77245496-77245496 on Build GRCh38
rs150358857 Chr.11: 77245411-77245411 on Build GRCh38
rs375831006 Chr.11: 77245250-77245250 on Build GRCh38
rs560582923 Chr.11: 77245548-77245550 on Build GRCh38
rs184766893 Chr.11: 77245331-77245331 on Build GRCh38
rs768126999 Chr.11: 77245266-77245266 on Build GRCh38

View Full Product Details