Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77918713-77919225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
UniGene
Hs.533723
Gene Chromosome Location
Chr.11: 77874491-77994849 on Build GRCh38
Transcripts
NM_033547.3 XM_011545352.2 XM_017018560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828001 c.1839C>T p.F613F Chr.11: 77918904-77918904 on Build GRCh38
7372329 c.1802C>G p.P601R Chr.11: 77918941-77918941 on Build GRCh38
6310156 c.1799C>G p.S600C Chr.11: 77918944-77918944 on Build GRCh38
161751 c.1771G>A p.G591S Chr.11: 77918972-77918972 on Build GRCh38
4532071 c.1850G>A p.S617N Chr.11: 77918893-77918893 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567637015 Chr.11: 77919067-77919067 on Build GRCh38
rs934889617 Chr.11: 77918881-77918881 on Build GRCh38
rs4084253 Chr.11: 77918913-77918913 on Build GRCh38
rs1047691613 Chr.11: 77919039-77919039 on Build GRCh38
rs764314282 Chr.11: 77918892-77918892 on Build GRCh38
rs537528042 Chr.11: 77918984-77918984 on Build GRCh38
rs750488651 Chr.11: 77918787-77918787 on Build GRCh38
rs756207239 Chr.11: 77918953-77918953 on Build GRCh38
rs766715145 Chr.11: 77918969-77918969 on Build GRCh38
rs551023724 Chr.11: 77919021-77919021 on Build GRCh38
rs540170965 Chr.11: 77919101-77919101 on Build GRCh38
rs774758528 Chr.11: 77918903-77918903 on Build GRCh38
rs772662807 Chr.11: 77918859-77918859 on Build GRCh38
rs770580492 Chr.11: 77918878-77918878 on Build GRCh38
rs536962646 Chr.11: 77919090-77919090 on Build GRCh38
rs571689070 Chr.11: 77918851-77918851 on Build GRCh38
rs758323244 Chr.11: 77919011-77919011 on Build GRCh38
rs754073240 Chr.11: 77918973-77918973 on Build GRCh38
rs4084254 Chr.11: 77918783-77918783 on Build GRCh38
rs1035621101 Chr.11: 77919086-77919086 on Build GRCh38
rs553352625 Chr.11: 77919169-77919169 on Build GRCh38
rs765934293 Chr.11: 77919008-77919009 on Build GRCh38
rs778612336 Chr.11: 77919000-77919000 on Build GRCh38
rs35582908 Chr.11: 77919023-77919024 on Build GRCh38
rs199923687 Chr.11: 77919140-77919141 on Build GRCh38
rs763334609 Chr.11: 77918889-77918889 on Build GRCh38
rs750512604 Chr.11: 77919102-77919102 on Build GRCh38
rs1039198819 Chr.11: 77918975-77918975 on Build GRCh38
rs748084985 Chr.11: 77918830-77918830 on Build GRCh38
rs1004111471 Chr.11: 77919066-77919066 on Build GRCh38
rs778941867 Chr.11: 77918825-77918825 on Build GRCh38
rs769332604 Chr.11: 77918805-77918805 on Build GRCh38
rs1023260138 Chr.11: 77919151-77919156 on Build GRCh38
rs886443476 Chr.11: 77919049-77919049 on Build GRCh38
rs772562430 Chr.11: 77918813-77918817 on Build GRCh38
rs756475119 Chr.11: 77918795-77918795 on Build GRCh38
rs112286182 Chr.11: 77918815-77918815 on Build GRCh38
rs751966254 Chr.11: 77918774-77918774 on Build GRCh38
rs747969884 Chr.11: 77919004-77919004 on Build GRCh38
rs968462411 Chr.11: 77919160-77919160 on Build GRCh38
rs1012737924 Chr.11: 77919132-77919132 on Build GRCh38
rs750591754 Chr.11: 77918925-77918925 on Build GRCh38
rs368882539 Chr.11: 77918870-77918870 on Build GRCh38
rs762587169 Chr.11: 77918998-77918999 on Build GRCh38
rs930754895 Chr.11: 77919033-77919033 on Build GRCh38
rs978920905 Chr.11: 77919166-77919166 on Build GRCh38
rs762271394 Chr.11: 77918916-77918916 on Build GRCh38
rs780411981 Chr.11: 77918800-77918800 on Build GRCh38
rs139356522 Chr.11: 77918834-77918834 on Build GRCh38
rs749110743 Chr.11: 77918805-77918805 on Build GRCh38
rs767547125 Chr.11: 77918778-77918778 on Build GRCh38
rs532161804 Chr.11: 77918745-77918745 on Build GRCh38
rs768066317 Chr.11: 77918924-77918924 on Build GRCh38
rs796677632 Chr.11: 77919095-77919095 on Build GRCh38
rs746508462 Chr.11: 77918877-77918877 on Build GRCh38

View Full Product Details