Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86303265-86303764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
UniGene
Hs.283322
Gene Chromosome Location
Chr.11: 86302211-86345943 on Build GRCh38
Transcripts
NM_001322404.1 NM_001322407.1 NM_001322409.1 NM_016401.3 NR_024597.1 NR_024598.1 NR_136324.1 XM_017017914.1 XM_017017915.1

SNP Detail

SNP ID Chromosome Location
rs187862846 Chr.11: 86303733-86303733 on Build GRCh38
rs1051247971 Chr.11: 86303433-86303433 on Build GRCh38
rs943365204 Chr.11: 86303727-86303727 on Build GRCh38
rs182638467 Chr.11: 86303311-86303311 on Build GRCh38
rs995382953 Chr.11: 86303546-86303546 on Build GRCh38
rs562677813 Chr.11: 86303642-86303642 on Build GRCh38
rs192536129 Chr.11: 86303553-86303553 on Build GRCh38
rs988623660 Chr.11: 86303321-86303321 on Build GRCh38
rs528955710 Chr.11: 86303483-86303483 on Build GRCh38
rs970260317 Chr.11: 86303587-86303587 on Build GRCh38
rs994078683 Chr.11: 86303504-86303504 on Build GRCh38
rs933462255 Chr.11: 86303717-86303717 on Build GRCh38
rs981626304 Chr.11: 86303302-86303302 on Build GRCh38
rs1026515417 Chr.11: 86303528-86303528 on Build GRCh38
rs914448585 Chr.11: 86303671-86303671 on Build GRCh38
rs889141778 Chr.11: 86303384-86303384 on Build GRCh38
rs1036644519 Chr.11: 86303516-86303516 on Build GRCh38
rs974957723 Chr.11: 86303687-86303687 on Build GRCh38
rs186684486 Chr.11: 86303448-86303448 on Build GRCh38
rs756950863 Chr.11: 86303652-86303652 on Build GRCh38
rs368940671 Chr.11: 86303493-86303493 on Build GRCh38
rs956076568 Chr.11: 86303654-86303654 on Build GRCh38
rs947605810 Chr.11: 86303335-86303335 on Build GRCh38
rs914797898 Chr.11: 86303332-86303332 on Build GRCh38
rs943362605 Chr.11: 86303394-86303394 on Build GRCh38
rs988839275 Chr.11: 86303662-86303662 on Build GRCh38
rs565422920 Chr.11: 86303308-86303308 on Build GRCh38
rs914482452 Chr.11: 86303338-86303338 on Build GRCh38
rs77569073 Chr.11: 86303643-86303643 on Build GRCh38
rs551588363 Chr.11: 86303689-86303689 on Build GRCh38
rs139953489 Chr.11: 86303352-86303352 on Build GRCh38
rs1049632307 Chr.11: 86303368-86303368 on Build GRCh38
rs955781656 Chr.11: 86303660-86303660 on Build GRCh38
rs1023208986 Chr.11: 86303610-86303610 on Build GRCh38
rs550098768 Chr.11: 86303589-86303589 on Build GRCh38
rs34104743 Chr.11: 86303492-86303492 on Build GRCh38
rs374522436 Chr.11: 86303468-86303468 on Build GRCh38
rs969693635 Chr.11: 86303290-86303290 on Build GRCh38
rs928659117 Chr.11: 86303306-86303307 on Build GRCh38
rs561325981 Chr.11: 86303462-86303462 on Build GRCh38
rs923254879 Chr.11: 86303735-86303735 on Build GRCh38
rs780768151 Chr.11: 86303665-86303665 on Build GRCh38
rs1039242319 Chr.11: 86303349-86303349 on Build GRCh38
rs184756606 Chr.11: 86303699-86303699 on Build GRCh38
rs968507178 Chr.11: 86303678-86303678 on Build GRCh38
rs932426735 Chr.11: 86303375-86303375 on Build GRCh38

View Full Product Details