Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86344870-86345395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
UniGene
Hs.283322
Gene Chromosome Location
Chr.11: 86302211-86345943 on Build GRCh38
Transcripts
NM_001322404.1 NM_001322407.1 NM_001322409.1 NM_016401.3 NR_024597.1 NR_024598.1 NR_136324.1 XM_017017914.1 XM_017017915.1

SNP Detail

SNP ID Chromosome Location
rs968737173 Chr.11: 86345188-86345188 on Build GRCh38
rs922199376 Chr.11: 86345222-86345222 on Build GRCh38
rs556395697 Chr.11: 86344946-86344946 on Build GRCh38
rs572016024 Chr.11: 86345219-86345219 on Build GRCh38
rs186219113 Chr.11: 86345291-86345291 on Build GRCh38
rs887828564 Chr.11: 86345070-86345070 on Build GRCh38
rs919614404 Chr.11: 86345292-86345292 on Build GRCh38
rs976639188 Chr.11: 86345274-86345274 on Build GRCh38
rs890893772 Chr.11: 86344952-86344952 on Build GRCh38
rs1036774499 Chr.11: 86345290-86345290 on Build GRCh38
rs898157221 Chr.11: 86345034-86345034 on Build GRCh38
rs976631205 Chr.11: 86345015-86345015 on Build GRCh38
rs563575596 Chr.11: 86345340-86345340 on Build GRCh38
rs577032059 Chr.11: 86345351-86345351 on Build GRCh38
rs987646847 Chr.11: 86345267-86345267 on Build GRCh38
rs181748900 Chr.11: 86345048-86345048 on Build GRCh38
rs985669164 Chr.11: 86344991-86344991 on Build GRCh38
rs80189718 Chr.11: 86345288-86345288 on Build GRCh38
rs538606812 Chr.11: 86345185-86345185 on Build GRCh38
rs954913715 Chr.11: 86345246-86345246 on Build GRCh38
rs1044479777 Chr.11: 86345358-86345358 on Build GRCh38
rs528472088 Chr.11: 86345365-86345365 on Build GRCh38
rs988598290 Chr.11: 86344909-86344909 on Build GRCh38
rs554749942 Chr.11: 86345280-86345280 on Build GRCh38
rs111497820 Chr.11: 86345281-86345281 on Build GRCh38
rs550853570 Chr.11: 86345107-86345107 on Build GRCh38
rs190680463 Chr.11: 86345357-86345357 on Build GRCh38
rs1009872344 Chr.11: 86345164-86345164 on Build GRCh38
rs147733341 Chr.11: 86345215-86345215 on Build GRCh38
rs943986553 Chr.11: 86345005-86345005 on Build GRCh38
rs891471211 Chr.11: 86345157-86345157 on Build GRCh38
rs10792848 Chr.11: 86345364-86345364 on Build GRCh38
rs1003162187 Chr.11: 86345124-86345124 on Build GRCh38
rs1047818012 Chr.11: 86345067-86345067 on Build GRCh38
rs1051911008 Chr.11: 86344932-86344932 on Build GRCh38
rs185831604 Chr.11: 86344927-86344927 on Build GRCh38
rs114235084 Chr.11: 86345118-86345118 on Build GRCh38
rs192292965 Chr.11: 86344992-86344992 on Build GRCh38
rs1057391538 Chr.11: 86345154-86345154 on Build GRCh38
rs1037107653 Chr.11: 86345032-86345032 on Build GRCh38
rs950730444 Chr.11: 86345312-86345312 on Build GRCh38
rs923314768 Chr.11: 86345046-86345046 on Build GRCh38
rs906019962 Chr.11: 86345120-86345120 on Build GRCh38
rs983832169 Chr.11: 86345332-86345332 on Build GRCh38
rs1035456885 Chr.11: 86345143-86345143 on Build GRCh38
rs942233025 Chr.11: 86345083-86345083 on Build GRCh38
rs534659008 Chr.11: 86345224-86345224 on Build GRCh38
rs965180659 Chr.11: 86345255-86345255 on Build GRCh38
rs1044305548 Chr.11: 86345082-86345082 on Build GRCh38
rs1021752924 Chr.11: 86344915-86344915 on Build GRCh38
rs905728358 Chr.11: 86345113-86345113 on Build GRCh38
rs775100177 Chr.11: 86345230-86345230 on Build GRCh38

View Full Product Details