Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 89691154-89691656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1B
Entrez Gene ID
219595
Gene Name
folate hydrolase 1B
Gene Aliases
FOLH2, FOLHP, GCP3, GCPIII, PSM, PSMA-LIKE, PSMAL
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 89637575-89698718 on Build GRCh38
Transcripts
NM_153696.2 XM_017017319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
691129 c.990T>A p.N330K Chr.11: 89691472-89691472 on Build GRCh38
5431898 c.1036G>C p.D346H Chr.11: 89691518-89691518 on Build GRCh38
691130 c.965-1G>T p.? Chr.11: 89691446-89691446 on Build GRCh38
1357670 c.988A>C p.N330H Chr.11: 89691470-89691470 on Build GRCh38
3454216 c.1026C>T p.L342L Chr.11: 89691508-89691508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756838627 Chr.11: 89691578-89691578 on Build GRCh38
rs780412338 Chr.11: 89691579-89691579 on Build GRCh38
rs35589251 Chr.11: 89691571-89691571 on Build GRCh38
rs59370786 Chr.11: 89691570-89691593 on Build GRCh38
rs778566402 Chr.11: 89691569-89691588 on Build GRCh38
rs753864777 Chr.11: 89691564-89691564 on Build GRCh38
rs199913180 Chr.11: 89691474-89691474 on Build GRCh38
rs755929474 Chr.11: 89691180-89691180 on Build GRCh38
rs878989493 Chr.11: 89691571-89691592 on Build GRCh38
rs979132331 Chr.11: 89691295-89691295 on Build GRCh38
rs753540097 Chr.11: 89691465-89691465 on Build GRCh38
rs924739190 Chr.11: 89691296-89691296 on Build GRCh38
rs747628722 Chr.11: 89691413-89691413 on Build GRCh38
rs746429533 Chr.11: 89691423-89691423 on Build GRCh38
rs371598284 Chr.11: 89691530-89691530 on Build GRCh38
rs968698427 Chr.11: 89691259-89691259 on Build GRCh38
rs68122964 Chr.11: 89691615-89691616 on Build GRCh38
rs773303060 Chr.11: 89691573-89691586 on Build GRCh38
rs752408664 Chr.11: 89691494-89691494 on Build GRCh38
rs368769742 Chr.11: 89691380-89691381 on Build GRCh38
rs35319604 Chr.11: 89691196-89691197 on Build GRCh38
rs761969715 Chr.11: 89691450-89691450 on Build GRCh38
rs766359131 Chr.11: 89691464-89691464 on Build GRCh38
rs548969559 Chr.11: 89691502-89691502 on Build GRCh38
rs778570844 Chr.11: 89691402-89691402 on Build GRCh38
rs765370576 Chr.11: 89691561-89691562 on Build GRCh38
rs1044955641 Chr.11: 89691441-89691441 on Build GRCh38
rs529230787 Chr.11: 89691500-89691500 on Build GRCh38
rs2035677 Chr.11: 89691548-89691548 on Build GRCh38
rs561763100 Chr.11: 89691207-89691207 on Build GRCh38
rs372213135 Chr.11: 89691466-89691466 on Build GRCh38
rs766932237 Chr.11: 89691569-89691602 on Build GRCh38
rs2130024 Chr.11: 89691572-89691572 on Build GRCh38
rs2130023 Chr.11: 89691570-89691570 on Build GRCh38
rs373487838 Chr.11: 89691455-89691455 on Build GRCh38
rs934712382 Chr.11: 89691338-89691338 on Build GRCh38
rs192957620 Chr.11: 89691634-89691634 on Build GRCh38
rs975643792 Chr.11: 89691241-89691241 on Build GRCh38
rs751001699 Chr.11: 89691577-89691577 on Build GRCh38
rs771157801 Chr.11: 89691573-89691588 on Build GRCh38
rs763440811 Chr.11: 89691433-89691433 on Build GRCh38
rs553576600 Chr.11: 89691596-89691596 on Build GRCh38
rs375039100 Chr.11: 89691304-89691304 on Build GRCh38
rs773358476 Chr.11: 89691560-89691561 on Build GRCh38
rs551107413 Chr.11: 89691562-89691562 on Build GRCh38
rs759317067 Chr.11: 89691569-89691569 on Build GRCh38
rs71472260 Chr.11: 89691570-89691571 on Build GRCh38
rs762057186 Chr.11: 89691547-89691547 on Build GRCh38
rs573346847 Chr.11: 89691598-89691598 on Build GRCh38
rs200000727 Chr.11: 89691569-89691594 on Build GRCh38
rs372697843 Chr.11: 89691270-89691270 on Build GRCh38
rs753677596 Chr.11: 89691187-89691187 on Build GRCh38
rs781717558 Chr.11: 89691422-89691422 on Build GRCh38
rs746786228 Chr.11: 89691571-89691586 on Build GRCh38
rs376902154 Chr.11: 89691461-89691461 on Build GRCh38
rs767768923 Chr.11: 89691453-89691453 on Build GRCh38
rs745456572 Chr.11: 89691569-89691586 on Build GRCh38
rs752722014 Chr.11: 89691568-89691569 on Build GRCh38
rs201929801 Chr.11: 89691592-89691592 on Build GRCh38
rs866294622 Chr.11: 89691616-89691616 on Build GRCh38
rs775660808 Chr.11: 89691427-89691427 on Build GRCh38
rs761857569 Chr.11: 89691561-89691564 on Build GRCh38
rs760837086 Chr.11: 89691558-89691558 on Build GRCh38
rs768048070 Chr.11: 89691569-89691590 on Build GRCh38
rs765086800 Chr.11: 89691571-89691571 on Build GRCh38
rs757900854 Chr.11: 89691499-89691499 on Build GRCh38
rs779674452 Chr.11: 89691571-89691588 on Build GRCh38
rs35123461 Chr.11: 89691573-89691573 on Build GRCh38
rs768383617 Chr.11: 89691571-89691600 on Build GRCh38
rs754810042 Chr.11: 89691472-89691472 on Build GRCh38
rs563805057 Chr.11: 89691425-89691425 on Build GRCh38
rs771623056 Chr.11: 89691419-89691419 on Build GRCh38
rs182573044 Chr.11: 89691194-89691194 on Build GRCh38
rs769807740 Chr.11: 89691573-89691590 on Build GRCh38
rs539523692 Chr.11: 89691574-89691574 on Build GRCh38
rs763830715 Chr.11: 89691576-89691576 on Build GRCh38
rs546803493 Chr.11: 89691447-89691447 on Build GRCh38
rs965646604 Chr.11: 89691223-89691223 on Build GRCh38
rs770151811 Chr.11: 89691504-89691504 on Build GRCh38
rs543905843 Chr.11: 89691300-89691300 on Build GRCh38
rs774488878 Chr.11: 89691581-89691616 on Build GRCh38
rs768669258 Chr.11: 89691479-89691480 on Build GRCh38
rs763176296 Chr.11: 89691561-89691566 on Build GRCh38
rs762820339 Chr.11: 89691573-89691592 on Build GRCh38
rs188596618 Chr.11: 89691444-89691444 on Build GRCh38
rs779643189 Chr.11: 89691183-89691183 on Build GRCh38
rs773114105 Chr.11: 89691550-89691550 on Build GRCh38
rs757988780 Chr.11: 89691575-89691575 on Build GRCh38
rs988021642 Chr.11: 89691365-89691365 on Build GRCh38
rs6144434 Chr.11: 89691569-89691592 on Build GRCh38
rs769199915 Chr.11: 89691521-89691521 on Build GRCh38
rs756820739 Chr.11: 89691569-89691596 on Build GRCh38
rs371051190 Chr.11: 89691512-89691512 on Build GRCh38
rs763125399 Chr.11: 89691573-89691598 on Build GRCh38
rs377645225 Chr.11: 89691570-89691570 on Build GRCh38
rs758146542 Chr.11: 89691571-89691590 on Build GRCh38
rs1028365660 Chr.11: 89691252-89691252 on Build GRCh38
rs776471833 Chr.11: 89691557-89691558 on Build GRCh38

View Full Product Details