Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92697304-92697802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017185.1 XM_017017186.1 XM_017017187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367689 c.3622A>C p.T1208P Chr.11: 92697398-92697398 on Build GRCh38
1357738 c.3640C>T p.R1214* Chr.11: 92697416-92697416 on Build GRCh38
133514 c.3663_3664TC>CA p.L1222M Chr.11: 92697439-92697440 on Build GRCh38
7250495 c.3612G>A p.L1204L Chr.11: 92697388-92697388 on Build GRCh38
4037514 c.3629G>T p.R1210M Chr.11: 92697405-92697405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767581941 Chr.11: 92697474-92697474 on Build GRCh38
rs3911560 Chr.11: 92697557-92697557 on Build GRCh38
rs746550104 Chr.11: 92697434-92697434 on Build GRCh38
rs955837249 Chr.11: 92697512-92697512 on Build GRCh38
rs779901514 Chr.11: 92697361-92697361 on Build GRCh38
rs754830139 Chr.11: 92697386-92697386 on Build GRCh38
rs1051756617 Chr.11: 92697355-92697355 on Build GRCh38
rs72958588 Chr.11: 92697555-92697555 on Build GRCh38
rs773458251 Chr.11: 92697480-92697480 on Build GRCh38
rs988984361 Chr.11: 92697533-92697533 on Build GRCh38
rs770444566 Chr.11: 92697451-92697451 on Build GRCh38
rs753780421 Chr.11: 92697774-92697774 on Build GRCh38
rs986969275 Chr.11: 92697718-92697718 on Build GRCh38
rs112729684 Chr.11: 92697665-92697665 on Build GRCh38
rs574094603 Chr.11: 92697762-92697762 on Build GRCh38
rs971334977 Chr.11: 92697606-92697606 on Build GRCh38
rs766313260 Chr.11: 92697768-92697768 on Build GRCh38
rs998069748 Chr.11: 92697417-92697417 on Build GRCh38
rs962440715 Chr.11: 92697683-92697683 on Build GRCh38
rs555969883 Chr.11: 92697675-92697675 on Build GRCh38
rs1010055907 Chr.11: 92697578-92697578 on Build GRCh38
rs901021002 Chr.11: 92697410-92697410 on Build GRCh38
rs368219689 Chr.11: 92697456-92697456 on Build GRCh38
rs973771882 Chr.11: 92697688-92697688 on Build GRCh38
rs79390114 Chr.11: 92697496-92697496 on Build GRCh38
rs368518991 Chr.11: 92697421-92697421 on Build GRCh38
rs563772749 Chr.11: 92697607-92697607 on Build GRCh38
rs760736205 Chr.11: 92697482-92697482 on Build GRCh38
rs559947043 Chr.11: 92697528-92697528 on Build GRCh38
rs866633555 Chr.11: 92697443-92697443 on Build GRCh38
rs749537814 Chr.11: 92697602-92697602 on Build GRCh38
rs770305020 Chr.11: 92697352-92697352 on Build GRCh38
rs376289468 Chr.11: 92697374-92697374 on Build GRCh38
rs113735851 Chr.11: 92697559-92697559 on Build GRCh38
rs753774128 Chr.11: 92697371-92697371 on Build GRCh38
rs998059999 Chr.11: 92697741-92697745 on Build GRCh38
rs927588750 Chr.11: 92697672-92697672 on Build GRCh38
rs370180105 Chr.11: 92697360-92697360 on Build GRCh38
rs747806208 Chr.11: 92697424-92697424 on Build GRCh38
rs1028239350 Chr.11: 92697761-92697761 on Build GRCh38
rs555830974 Chr.11: 92697712-92697712 on Build GRCh38
rs774788087 Chr.11: 92697459-92697459 on Build GRCh38
rs4122086 Chr.11: 92697680-92697680 on Build GRCh38
rs779963435 Chr.11: 92697355-92697356 on Build GRCh38
rs1024663343 Chr.11: 92697604-92697604 on Build GRCh38
rs1032654931 Chr.11: 92697770-92697770 on Build GRCh38
rs750665793 Chr.11: 92697511-92697511 on Build GRCh38
rs186569910 Chr.11: 92697765-92697765 on Build GRCh38
rs561638914 Chr.11: 92697719-92697719 on Build GRCh38
rs371158510 Chr.11: 92697547-92697547 on Build GRCh38
rs140155860 Chr.11: 92697722-92697722 on Build GRCh38
rs574141235 Chr.11: 92697328-92697328 on Build GRCh38
rs925250478 Chr.11: 92697775-92697775 on Build GRCh38
rs930165839 Chr.11: 92697713-92697713 on Build GRCh38
rs772381927 Chr.11: 92697614-92697614 on Build GRCh38
rs372525516 Chr.11: 92697432-92697432 on Build GRCh38
rs1039724450 Chr.11: 92697454-92697454 on Build GRCh38
rs762123364 Chr.11: 92697466-92697466 on Build GRCh38
rs377092202 Chr.11: 92697452-92697452 on Build GRCh38
rs773269671 Chr.11: 92697668-92697668 on Build GRCh38
rs766437342 Chr.11: 92697489-92697489 on Build GRCh38

View Full Product Details