Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93499198-93499706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO4
Entrez Gene ID
56935
Gene Name
single-pass membrane protein with coiled-coil domains 4
Gene Aliases
C11orf75, FN5
UniGene
Hs.438064
Gene Chromosome Location
Chr.11: 93478472-93586856 on Build GRCh38
Transcripts
NM_020179.2 XM_011542907.1 XM_011542908.2 XM_011542909.1 XM_011542910.2 XM_011542911.2 XM_017018019.1 XM_017018020.1

SNP Detail

SNP ID Chromosome Location
rs1054844296 Chr.11: 93499602-93499602 on Build GRCh38
rs1033533002 Chr.11: 93499262-93499262 on Build GRCh38
rs756369726 Chr.11: 93499638-93499638 on Build GRCh38
rs530266850 Chr.11: 93499429-93499429 on Build GRCh38
rs997897739 Chr.11: 93499662-93499662 on Build GRCh38
rs575379276 Chr.11: 93499576-93499576 on Build GRCh38
rs538944723 Chr.11: 93499555-93499555 on Build GRCh38
rs192594506 Chr.11: 93499681-93499681 on Build GRCh38
rs77868322 Chr.11: 93499574-93499574 on Build GRCh38
rs11540635 Chr.11: 93499324-93499324 on Build GRCh38
rs976781522 Chr.11: 93499454-93499454 on Build GRCh38
rs183179434 Chr.11: 93499251-93499251 on Build GRCh38
rs1048867306 Chr.11: 93499670-93499670 on Build GRCh38
rs374643201 Chr.11: 93499495-93499495 on Build GRCh38
rs190996629 Chr.11: 93499459-93499459 on Build GRCh38
rs2605614 Chr.11: 93499579-93499579 on Build GRCh38
rs187713280 Chr.11: 93499550-93499550 on Build GRCh38
rs925908358 Chr.11: 93499346-93499346 on Build GRCh38
rs978643765 Chr.11: 93499333-93499333 on Build GRCh38
rs1046467266 Chr.11: 93499571-93499571 on Build GRCh38
rs921097255 Chr.11: 93499327-93499327 on Build GRCh38
rs965857759 Chr.11: 93499386-93499386 on Build GRCh38
rs182613780 Chr.11: 93499526-93499526 on Build GRCh38
rs953863706 Chr.11: 93499328-93499328 on Build GRCh38
rs796656412 Chr.11: 93499683-93499683 on Build GRCh38
rs11020382 Chr.11: 93499298-93499298 on Build GRCh38
rs915504288 Chr.11: 93499528-93499528 on Build GRCh38
rs927630732 Chr.11: 93499578-93499578 on Build GRCh38
rs567416547 Chr.11: 93499496-93499496 on Build GRCh38
rs554637991 Chr.11: 93499627-93499627 on Build GRCh38
rs1022057946 Chr.11: 93499363-93499363 on Build GRCh38
rs954246099 Chr.11: 93499520-93499520 on Build GRCh38
rs867005359 Chr.11: 93499605-93499605 on Build GRCh38
rs542515137 Chr.11: 93499437-93499437 on Build GRCh38
rs994762150 Chr.11: 93499293-93499293 on Build GRCh38
rs532847317 Chr.11: 93499368-93499368 on Build GRCh38
rs894939399 Chr.11: 93499269-93499269 on Build GRCh38
rs1027605951 Chr.11: 93499297-93499297 on Build GRCh38
rs965142259 Chr.11: 93499295-93499295 on Build GRCh38
rs888683492 Chr.11: 93499588-93499588 on Build GRCh38
rs1003899083 Chr.11: 93499270-93499270 on Build GRCh38
rs945637440 Chr.11: 93499538-93499538 on Build GRCh38
rs537576879 Chr.11: 93499666-93499666 on Build GRCh38
rs540276038 Chr.11: 93499280-93499280 on Build GRCh38
rs563079851 Chr.11: 93499378-93499378 on Build GRCh38
rs534552545 Chr.11: 93499497-93499497 on Build GRCh38
rs983210379 Chr.11: 93499352-93499352 on Build GRCh38
rs188168596 Chr.11: 93499374-93499374 on Build GRCh38

View Full Product Details