Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94082334-94082818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
-
UniGene
Hs.669645
Gene Chromosome Location
Chr.11: 94021212-94114208 on Build GRCh38
Transcripts
NM_001098672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6567801 c.1834G>A p.D612N Chr.11: 94082535-94082535 on Build GRCh38
933453 c.1858C>T p.R620* Chr.11: 94082559-94082559 on Build GRCh38
1209539 c.1802T>G p.F601C Chr.11: 94082503-94082503 on Build GRCh38
2061147 c.1859G>A p.R620Q Chr.11: 94082560-94082560 on Build GRCh38
933452 c.1764G>T p.E588D Chr.11: 94082465-94082465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376150745 Chr.11: 94082413-94082413 on Build GRCh38
rs144472999 Chr.11: 94082744-94082744 on Build GRCh38
rs1056347628 Chr.11: 94082425-94082425 on Build GRCh38
rs960176530 Chr.11: 94082560-94082560 on Build GRCh38
rs749596426 Chr.11: 94082384-94082384 on Build GRCh38
rs944494136 Chr.11: 94082718-94082719 on Build GRCh38
rs191008287 Chr.11: 94082512-94082512 on Build GRCh38
rs372167793 Chr.11: 94082700-94082700 on Build GRCh38
rs955613140 Chr.11: 94082633-94082633 on Build GRCh38
rs182720640 Chr.11: 94082572-94082572 on Build GRCh38
rs768714491 Chr.11: 94082590-94082590 on Build GRCh38
rs758014551 Chr.11: 94082736-94082736 on Build GRCh38
rs761835440 Chr.11: 94082505-94082505 on Build GRCh38
rs767378933 Chr.11: 94082375-94082375 on Build GRCh38
rs747552754 Chr.11: 94082437-94082437 on Build GRCh38
rs760542148 Chr.11: 94082533-94082533 on Build GRCh38
rs775680244 Chr.11: 94082481-94082481 on Build GRCh38
rs764831658 Chr.11: 94082554-94082554 on Build GRCh38
rs750134666 Chr.11: 94082378-94082378 on Build GRCh38
rs767898751 Chr.11: 94082444-94082444 on Build GRCh38
rs368679854 Chr.11: 94082453-94082453 on Build GRCh38
rs748144386 Chr.11: 94082615-94082615 on Build GRCh38
rs369174069 Chr.11: 94082528-94082528 on Build GRCh38
rs942769306 Chr.11: 94082575-94082575 on Build GRCh38
rs754517918 Chr.11: 94082547-94082547 on Build GRCh38
rs768787594 Chr.11: 94082492-94082492 on Build GRCh38
rs778225760 Chr.11: 94082435-94082435 on Build GRCh38
rs139764817 Chr.11: 94082588-94082588 on Build GRCh38
rs763104825 Chr.11: 94082484-94082484 on Build GRCh38
rs751638993 Chr.11: 94082450-94082451 on Build GRCh38
rs772837071 Chr.11: 94082530-94082532 on Build GRCh38
rs761444838 Chr.11: 94082589-94082589 on Build GRCh38
rs746204050 Chr.11: 94082469-94082469 on Build GRCh38
rs766133811 Chr.11: 94082788-94082788 on Build GRCh38
rs780331616 Chr.11: 94082587-94082587 on Build GRCh38
rs187290438 Chr.11: 94082735-94082735 on Build GRCh38
rs577714289 Chr.11: 94082545-94082545 on Build GRCh38
rs766188707 Chr.11: 94082534-94082534 on Build GRCh38
rs201293987 Chr.11: 94082394-94082394 on Build GRCh38
rs756484899 Chr.11: 94082586-94082586 on Build GRCh38
rs774256979 Chr.11: 94082499-94082499 on Build GRCh38
rs774537418 Chr.11: 94082608-94082608 on Build GRCh38
rs979510165 Chr.11: 94082777-94082777 on Build GRCh38
rs760028675 Chr.11: 94082557-94082557 on Build GRCh38
rs546931057 Chr.11: 94082759-94082759 on Build GRCh38
rs752127877 Chr.11: 94082559-94082559 on Build GRCh38
rs757824964 Chr.11: 94082569-94082569 on Build GRCh38
rs563428534 Chr.11: 94082582-94082582 on Build GRCh38
rs946789530 Chr.11: 94082369-94082369 on Build GRCh38
rs911745684 Chr.11: 94082704-94082704 on Build GRCh38
rs774582814 Chr.11: 94082423-94082423 on Build GRCh38
rs1023168244 Chr.11: 94082581-94082581 on Build GRCh38
rs1038873483 Chr.11: 94082721-94082724 on Build GRCh38
rs755879210 Chr.11: 94082380-94082380 on Build GRCh38
rs781659074 Chr.11: 94082462-94082462 on Build GRCh38
rs118179898 Chr.11: 94082624-94082624 on Build GRCh38
rs921388658 Chr.11: 94082739-94082739 on Build GRCh38
rs186352378 Chr.11: 94082371-94082371 on Build GRCh38

View Full Product Details