Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94545310-94545807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT4
Entrez Gene ID
2526
Gene Name
fucosyltransferase 4
Gene Aliases
CD15, ELFT, FCT3A, FUC-TIV, FUTIV, LeX, SSEA-1
UniGene
Hs.390420
Gene Chromosome Location
Chr.11: 94543851-94549898 on Build GRCh38
Transcripts
NM_002033.3 NR_135093.1

Gene Symbol
LOC105369438
Entrez Gene ID
105369438
Gene Name
uncharacterized LOC105369438
Gene Aliases
-
UniGene
Hs.568921
Gene Chromosome Location
Chr.11: 94545332-94740355 on Build GRCh38
Transcripts
NM_002033.3 NR_135093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6563743 c.1310C>G p.P437R Chr.11: 94545443-94545443 on Build GRCh38
4574898 c.1302G>C p.G434G Chr.11: 94545435-94545435 on Build GRCh38
4037709 c.1401C>A p.S467S Chr.11: 94545534-94545534 on Build GRCh38
6519488 c.1421T>A p.F474Y Chr.11: 94545554-94545554 on Build GRCh38
430098 c.1414C>T p.L472L Chr.11: 94545547-94545547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994201497 Chr.11: 94545733-94545733 on Build GRCh38
rs761020875 Chr.11: 94545346-94545346 on Build GRCh38
rs376104557 Chr.11: 94545654-94545654 on Build GRCh38
rs1029605096 Chr.11: 94545513-94545513 on Build GRCh38
rs199657418 Chr.11: 94545534-94545534 on Build GRCh38
rs755525118 Chr.11: 94545672-94545672 on Build GRCh38
rs756937681 Chr.11: 94545623-94545623 on Build GRCh38
rs911129913 Chr.11: 94545577-94545577 on Build GRCh38
rs371160166 Chr.11: 94545673-94545673 on Build GRCh38
rs866761099 Chr.11: 94545487-94545487 on Build GRCh38
rs776657373 Chr.11: 94545649-94545649 on Build GRCh38
rs763913859 Chr.11: 94545384-94545384 on Build GRCh38
rs753366955 Chr.11: 94545759-94545759 on Build GRCh38
rs747813301 Chr.11: 94545436-94545436 on Build GRCh38
rs760085870 Chr.11: 94545747-94545747 on Build GRCh38
rs146378578 Chr.11: 94545559-94545559 on Build GRCh38
rs780702810 Chr.11: 94545407-94545407 on Build GRCh38
rs745722154 Chr.11: 94545632-94545632 on Build GRCh38
rs778333294 Chr.11: 94545430-94545430 on Build GRCh38
rs776942740 Chr.11: 94545582-94545582 on Build GRCh38
rs779120740 Chr.11: 94545388-94545388 on Build GRCh38
rs766957343 Chr.11: 94545662-94545662 on Build GRCh38
rs767047234 Chr.11: 94545606-94545606 on Build GRCh38
rs372130626 Chr.11: 94545375-94545375 on Build GRCh38
rs201757138 Chr.11: 94545563-94545563 on Build GRCh38
rs369821585 Chr.11: 94545709-94545709 on Build GRCh38
rs972814488 Chr.11: 94545450-94545450 on Build GRCh38
rs762096760 Chr.11: 94545650-94545650 on Build GRCh38
rs759533317 Chr.11: 94545362-94545362 on Build GRCh38
rs781326016 Chr.11: 94545711-94545711 on Build GRCh38
rs767600530 Chr.11: 94545478-94545478 on Build GRCh38
rs2230274 Chr.11: 94545547-94545547 on Build GRCh38
rs200442017 Chr.11: 94545501-94545501 on Build GRCh38
rs768802187 Chr.11: 94545648-94545648 on Build GRCh38
rs748608722 Chr.11: 94545335-94545335 on Build GRCh38
rs985872066 Chr.11: 94545773-94545773 on Build GRCh38
rs758129924 Chr.11: 94545782-94545782 on Build GRCh38
rs754883134 Chr.11: 94545357-94545357 on Build GRCh38
rs752764588 Chr.11: 94545448-94545463 on Build GRCh38
rs771438475 Chr.11: 94545738-94545738 on Build GRCh38
rs778677048 Chr.11: 94545630-94545630 on Build GRCh38
rs769562458 Chr.11: 94545444-94545444 on Build GRCh38
rs142877244 Chr.11: 94545681-94545681 on Build GRCh38
rs200642870 Chr.11: 94545613-94545613 on Build GRCh38
rs527908470 Chr.11: 94545781-94545781 on Build GRCh38
rs1016991795 Chr.11: 94545411-94545411 on Build GRCh38
rs756960454 Chr.11: 94545403-94545403 on Build GRCh38
rs139614833 Chr.11: 94545619-94545619 on Build GRCh38
rs777246164 Chr.11: 94545618-94545618 on Build GRCh38
rs767357012 Chr.11: 94545363-94545364 on Build GRCh38
rs745632244 Chr.11: 94545692-94545692 on Build GRCh38
rs765298288 Chr.11: 94545366-94545366 on Build GRCh38
rs553637824 Chr.11: 94545387-94545387 on Build GRCh38
rs982556499 Chr.11: 94545627-94545627 on Build GRCh38
rs750891847 Chr.11: 94545592-94545592 on Build GRCh38
rs761352922 Chr.11: 94545768-94545768 on Build GRCh38
rs151024645 Chr.11: 94545721-94545721 on Build GRCh38
rs911078757 Chr.11: 94545609-94545609 on Build GRCh38
rs955334999 Chr.11: 94545763-94545763 on Build GRCh38
rs756239638 Chr.11: 94545623-94545625 on Build GRCh38
rs774306679 Chr.11: 94545470-94545470 on Build GRCh38
rs759566244 Chr.11: 94545475-94545475 on Build GRCh38
rs771038846 Chr.11: 94545351-94545351 on Build GRCh38
rs748696976 Chr.11: 94545427-94545427 on Build GRCh38
rs780851322 Chr.11: 94545522-94545522 on Build GRCh38
rs758965093 Chr.11: 94545594-94545594 on Build GRCh38
rs771976264 Chr.11: 94545634-94545634 on Build GRCh38
rs370382431 Chr.11: 94545578-94545578 on Build GRCh38
rs751374579 Chr.11: 94545786-94545786 on Build GRCh38
rs779567545 Chr.11: 94545423-94545423 on Build GRCh38
rs770746563 Chr.11: 94545464-94545464 on Build GRCh38
rs774218767 Chr.11: 94545543-94545543 on Build GRCh38
rs778587119 Chr.11: 94545685-94545685 on Build GRCh38
rs779901132 Chr.11: 94545784-94545784 on Build GRCh38
rs377261715 Chr.11: 94545697-94545697 on Build GRCh38
rs772875106 Chr.11: 94545451-94545451 on Build GRCh38
rs767968575 Chr.11: 94545750-94545750 on Build GRCh38
rs769313697 Chr.11: 94545530-94545530 on Build GRCh38
rs750987299 Chr.11: 94545510-94545510 on Build GRCh38
rs559167321 Chr.11: 94545720-94545720 on Build GRCh38
rs760803629 Chr.11: 94545489-94545489 on Build GRCh38
rs1029741008 Chr.11: 94545739-94545739 on Build GRCh38
rs754477714 Chr.11: 94545511-94545511 on Build GRCh38
rs746978754 Chr.11: 94545707-94545707 on Build GRCh38
rs762859697 Chr.11: 94545462-94545462 on Build GRCh38
rs762834047 Chr.11: 94545380-94545380 on Build GRCh38
rs544932035 Chr.11: 94545740-94545740 on Build GRCh38
rs140199740 Chr.11: 94545492-94545492 on Build GRCh38
rs762032604 Chr.11: 94545584-94545584 on Build GRCh38
rs756891265 Chr.11: 94545683-94545683 on Build GRCh38
rs201504879 Chr.11: 94545637-94545637 on Build GRCh38
rs548859980 Chr.11: 94545682-94545682 on Build GRCh38
rs765708849 Chr.11: 94545587-94545587 on Build GRCh38
rs776833805 Chr.11: 94545360-94545360 on Build GRCh38
rs773673789 Chr.11: 94545340-94545340 on Build GRCh38
rs779521687 Chr.11: 94545504-94545504 on Build GRCh38
rs752134160 Chr.11: 94545670-94545670 on Build GRCh38
rs779805643 Chr.11: 94545701-94545701 on Build GRCh38
rs755685530 Chr.11: 94545617-94545617 on Build GRCh38
rs774656660 Chr.11: 94545746-94545746 on Build GRCh38
rs763456816 Chr.11: 94545661-94545661 on Build GRCh38
rs368516050 Chr.11: 94545338-94545338 on Build GRCh38
rs374018726 Chr.11: 94545417-94545417 on Build GRCh38
rs749000680 Chr.11: 94545538-94545538 on Build GRCh38
rs200047243 Chr.11: 94545486-94545486 on Build GRCh38
rs373912875 Chr.11: 94545527-94545527 on Build GRCh38
rs772677516 Chr.11: 94545429-94545429 on Build GRCh38
rs573031841 Chr.11: 94545636-94545636 on Build GRCh38
rs747020066 Chr.11: 94545639-94545639 on Build GRCh38
rs896153520 Chr.11: 94545653-94545653 on Build GRCh38
rs371954490 Chr.11: 94545540-94545540 on Build GRCh38
rs763364563 Chr.11: 94545732-94545732 on Build GRCh38
rs775535065 Chr.11: 94545573-94545573 on Build GRCh38
rs541825961 Chr.11: 94545778-94545778 on Build GRCh38
rs867516152 Chr.11: 94545572-94545572 on Build GRCh38
rs763667950 Chr.11: 94545677-94545677 on Build GRCh38
rs765618978 Chr.11: 94545651-94545651 on Build GRCh38
rs750024102 Chr.11: 94545409-94545409 on Build GRCh38

View Full Product Details