Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 96392821-96393327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JRKL
Entrez Gene ID
8690
Gene Name
JRK-like
Gene Aliases
HHMJG
UniGene
Hs.105940
Gene Chromosome Location
Chr.11: 96389994-96393563 on Build GRCh38
Transcripts
NM_001261833.1 NM_003772.3

SNP Detail

SNP ID Chromosome Location
rs11021571 Chr.11: 96393146-96393146 on Build GRCh38
rs909367154 Chr.11: 96393205-96393205 on Build GRCh38
rs893010291 Chr.11: 96393018-96393021 on Build GRCh38
rs577389382 Chr.11: 96393301-96393301 on Build GRCh38
rs4753751 Chr.11: 96393197-96393197 on Build GRCh38
rs998812777 Chr.11: 96392972-96392972 on Build GRCh38
rs939826200 Chr.11: 96392875-96392875 on Build GRCh38
rs903091330 Chr.11: 96392960-96392960 on Build GRCh38
rs760903212 Chr.11: 96393125-96393126 on Build GRCh38
rs756971850 Chr.11: 96393089-96393089 on Build GRCh38
rs1042603793 Chr.11: 96393073-96393073 on Build GRCh38
rs919636139 Chr.11: 96392985-96392985 on Build GRCh38
rs908399372 Chr.11: 96392874-96392874 on Build GRCh38
rs11021572 Chr.11: 96393148-96393148 on Build GRCh38
rs555063849 Chr.11: 96393067-96393067 on Build GRCh38
rs866633290 Chr.11: 96393270-96393270 on Build GRCh38
rs559125732 Chr.11: 96393234-96393234 on Build GRCh38
rs768506912 Chr.11: 96393124-96393124 on Build GRCh38
rs971692139 Chr.11: 96392865-96392865 on Build GRCh38
rs1025372509 Chr.11: 96393227-96393227 on Build GRCh38
rs971147297 Chr.11: 96393080-96393080 on Build GRCh38
rs778368792 Chr.11: 96393099-96393099 on Build GRCh38
rs933294801 Chr.11: 96392928-96392928 on Build GRCh38
rs1049829982 Chr.11: 96393007-96393007 on Build GRCh38
rs985094037 Chr.11: 96393203-96393203 on Build GRCh38
rs200299383 Chr.11: 96393122-96393123 on Build GRCh38
rs11549459 Chr.11: 96393001-96393001 on Build GRCh38
rs761834872 Chr.11: 96393126-96393126 on Build GRCh38
rs201157332 Chr.11: 96393125-96393125 on Build GRCh38
rs181706691 Chr.11: 96393207-96393207 on Build GRCh38
rs981317451 Chr.11: 96392867-96392867 on Build GRCh38
rs767655092 Chr.11: 96393124-96393125 on Build GRCh38
rs575966360 Chr.11: 96392954-96392954 on Build GRCh38
rs549492797 Chr.11: 96393023-96393023 on Build GRCh38
rs1038163503 Chr.11: 96392958-96392958 on Build GRCh38
rs943438912 Chr.11: 96392880-96392880 on Build GRCh38
rs1053028 Chr.11: 96392989-96392989 on Build GRCh38
rs558403487 Chr.11: 96392907-96392907 on Build GRCh38
rs1022688783 Chr.11: 96393068-96393068 on Build GRCh38
rs751982571 Chr.11: 96392936-96392936 on Build GRCh38
rs35254662 Chr.11: 96393285-96393286 on Build GRCh38
rs902724950 Chr.11: 96393083-96393083 on Build GRCh38
rs959417135 Chr.11: 96393125-96393125 on Build GRCh38
rs1000987155 Chr.11: 96393093-96393093 on Build GRCh38

View Full Product Details