Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103054213-103054716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D5
Entrez Gene ID
84259
Gene Name
defective in cullin neddylation 1 domain containing 5
Gene Aliases
SCCRO5
UniGene
Hs.503716
Gene Chromosome Location
Chr.11: 103050684-103092215 on Build GRCh38
Transcripts
NM_001318739.1 NM_001318740.1 NM_001318741.1 NM_032299.3

SNP Detail

SNP ID Chromosome Location
rs1012586056 Chr.11: 103054509-103054509 on Build GRCh38
rs1009720019 Chr.11: 103054491-103054491 on Build GRCh38
rs185470582 Chr.11: 103054537-103054537 on Build GRCh38
rs555488014 Chr.11: 103054275-103054275 on Build GRCh38
rs1000096707 Chr.11: 103054619-103054619 on Build GRCh38
rs896476525 Chr.11: 103054383-103054383 on Build GRCh38
rs143763194 Chr.11: 103054318-103054318 on Build GRCh38
rs147189179 Chr.11: 103054628-103054628 on Build GRCh38
rs545559423 Chr.11: 103054474-103054474 on Build GRCh38
rs561672786 Chr.11: 103054605-103054605 on Build GRCh38
rs1053965247 Chr.11: 103054470-103054470 on Build GRCh38
rs779519724 Chr.11: 103054622-103054622 on Build GRCh38
rs940703743 Chr.11: 103054348-103054348 on Build GRCh38
rs909269455 Chr.11: 103054326-103054326 on Build GRCh38
rs755720335 Chr.11: 103054464-103054464 on Build GRCh38
rs554037792 Chr.11: 103054385-103054385 on Build GRCh38
rs535452295 Chr.11: 103054612-103054614 on Build GRCh38
rs1001012468 Chr.11: 103054440-103054440 on Build GRCh38
rs898024412 Chr.11: 103054461-103054461 on Build GRCh38
rs1031537278 Chr.11: 103054309-103054309 on Build GRCh38
rs575259454 Chr.11: 103054309-103054309 on Build GRCh38
rs904189323 Chr.11: 103054616-103054617 on Build GRCh38
rs116852709 Chr.11: 103054629-103054629 on Build GRCh38
rs1012556674 Chr.11: 103054248-103054248 on Build GRCh38
rs1019326829 Chr.11: 103054507-103054507 on Build GRCh38
rs972114896 Chr.11: 103054631-103054631 on Build GRCh38
rs1038353194 Chr.11: 103054451-103054451 on Build GRCh38
rs1045287051 Chr.11: 103054402-103054402 on Build GRCh38
rs968614273 Chr.11: 103054289-103054289 on Build GRCh38
rs996771131 Chr.11: 103054581-103054581 on Build GRCh38
rs180867830 Chr.11: 103054418-103054418 on Build GRCh38
rs929447861 Chr.11: 103054472-103054472 on Build GRCh38
rs753225347 Chr.11: 103054384-103054384 on Build GRCh38
rs977003137 Chr.11: 103054661-103054661 on Build GRCh38
rs771692448 Chr.11: 103054316-103054316 on Build GRCh38
rs895507974 Chr.11: 103054235-103054235 on Build GRCh38
rs931093000 Chr.11: 103054313-103054313 on Build GRCh38
rs754537293 Chr.11: 103054606-103054606 on Build GRCh38
rs80119485 Chr.11: 103054349-103054349 on Build GRCh38
rs965041837 Chr.11: 103054555-103054555 on Build GRCh38
rs570528006 Chr.11: 103054693-103054693 on Build GRCh38
rs975322021 Chr.11: 103054278-103054278 on Build GRCh38
rs531323176 Chr.11: 103054531-103054531 on Build GRCh38
rs910518402 Chr.11: 103054436-103054436 on Build GRCh38
rs562606524 Chr.11: 103054475-103054475 on Build GRCh38
rs766056056 Chr.11: 103054293-103054293 on Build GRCh38
rs1028208318 Chr.11: 103054602-103054602 on Build GRCh38
rs933361995 Chr.11: 103054398-103054399 on Build GRCh38

View Full Product Details