Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103057269-103057810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D5
Entrez Gene ID
84259
Gene Name
defective in cullin neddylation 1 domain containing 5
Gene Aliases
SCCRO5
UniGene
Hs.503716
Gene Chromosome Location
Chr.11: 103050684-103092215 on Build GRCh38
Transcripts
NM_001318739.1 NM_001318740.1 NM_001318741.1 NM_032299.3

SNP Detail

SNP ID Chromosome Location
rs557880778 Chr.11: 103057471-103057471 on Build GRCh38
rs986893523 Chr.11: 103057587-103057587 on Build GRCh38
rs966995098 Chr.11: 103057776-103057776 on Build GRCh38
rs2509111 Chr.11: 103057721-103057721 on Build GRCh38
rs146846392 Chr.11: 103057522-103057522 on Build GRCh38
rs907389650 Chr.11: 103057674-103057674 on Build GRCh38
rs188840421 Chr.11: 103057555-103057555 on Build GRCh38
rs1055970676 Chr.11: 103057698-103057698 on Build GRCh38
rs1020745879 Chr.11: 103057721-103057721 on Build GRCh38
rs566155672 Chr.11: 103057613-103057613 on Build GRCh38
rs977333066 Chr.11: 103057537-103057537 on Build GRCh38
rs796198532 Chr.11: 103057377-103057377 on Build GRCh38
rs954493106 Chr.11: 103057578-103057578 on Build GRCh38
rs1024563937 Chr.11: 103057341-103057341 on Build GRCh38
rs914653706 Chr.11: 103057419-103057419 on Build GRCh38
rs185355130 Chr.11: 103057701-103057701 on Build GRCh38
rs145371778 Chr.11: 103057768-103057769 on Build GRCh38
rs912594327 Chr.11: 103057779-103057779 on Build GRCh38
rs75039344 Chr.11: 103057552-103057552 on Build GRCh38
rs970624136 Chr.11: 103057347-103057348 on Build GRCh38
rs568245892 Chr.11: 103057444-103057445 on Build GRCh38
rs990275551 Chr.11: 103057397-103057397 on Build GRCh38
rs117674362 Chr.11: 103057675-103057675 on Build GRCh38
rs1047266145 Chr.11: 103057669-103057669 on Build GRCh38
rs549945946 Chr.11: 103057465-103057465 on Build GRCh38
rs1033507723 Chr.11: 103057392-103057392 on Build GRCh38
rs542524336 Chr.11: 103057747-103057747 on Build GRCh38
rs942758603 Chr.11: 103057616-103057616 on Build GRCh38
rs749665426 Chr.11: 103057502-103057502 on Build GRCh38
rs200708074 Chr.11: 103057554-103057555 on Build GRCh38
rs550795781 Chr.11: 103057709-103057709 on Build GRCh38
rs1033540481 Chr.11: 103057676-103057676 on Build GRCh38
rs556835413 Chr.11: 103057731-103057731 on Build GRCh38
rs75794670 Chr.11: 103057719-103057719 on Build GRCh38
rs749281699 Chr.11: 103057702-103057703 on Build GRCh38
rs930269936 Chr.11: 103057667-103057667 on Build GRCh38
rs1011217877 Chr.11: 103057403-103057403 on Build GRCh38
rs1021329890 Chr.11: 103057744-103057744 on Build GRCh38
rs541771590 Chr.11: 103057623-103057623 on Build GRCh38
rs536688053 Chr.11: 103057714-103057714 on Build GRCh38
rs537534617 Chr.11: 103057424-103057424 on Build GRCh38
rs958629332 Chr.11: 103057348-103057348 on Build GRCh38
rs552963312 Chr.11: 103057494-103057494 on Build GRCh38
rs113032356 Chr.11: 103057552-103057553 on Build GRCh38
rs977494659 Chr.11: 103057737-103057737 on Build GRCh38
rs540070616 Chr.11: 103057551-103057552 on Build GRCh38
rs1048528131 Chr.11: 103057554-103057554 on Build GRCh38
rs533394245 Chr.11: 103057525-103057525 on Build GRCh38
rs757673535 Chr.11: 103057462-103057462 on Build GRCh38
rs561944756 Chr.11: 103057628-103057628 on Build GRCh38
rs550482337 Chr.11: 103057531-103057531 on Build GRCh38
rs1039963792 Chr.11: 103057507-103057507 on Build GRCh38
rs557596421 Chr.11: 103057691-103057691 on Build GRCh38
rs140639725 Chr.11: 103057735-103057735 on Build GRCh38
rs958098278 Chr.11: 103057396-103057396 on Build GRCh38
rs912563478 Chr.11: 103057482-103057482 on Build GRCh38
rs193030093 Chr.11: 103057594-103057594 on Build GRCh38
rs746533727 Chr.11: 103057350-103057350 on Build GRCh38

View Full Product Details