Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 109426393-109426898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf87
Entrez Gene ID
399947
Gene Name
chromosome 11 open reading frame 87
Gene Aliases
LOH11CR1A, NEURIM1
UniGene
Hs.172982
Gene Chromosome Location
Chr.11: 109422120-109433003 on Build GRCh38
Transcripts
NM_207645.3 XM_011542817.2 XM_011542818.2

SNP Detail

SNP ID Chromosome Location
rs377556482 Chr.11: 109426822-109426822 on Build GRCh38
rs534479814 Chr.11: 109426458-109426458 on Build GRCh38
rs998408459 Chr.11: 109426784-109426784 on Build GRCh38
rs547446506 Chr.11: 109426843-109426843 on Build GRCh38
rs866444636 Chr.11: 109426543-109426543 on Build GRCh38
rs117469332 Chr.11: 109426678-109426678 on Build GRCh38
rs535167917 Chr.11: 109426616-109426616 on Build GRCh38
rs149791407 Chr.11: 109426736-109426736 on Build GRCh38
rs1000636210 Chr.11: 109426699-109426699 on Build GRCh38
rs141007314 Chr.11: 109426501-109426501 on Build GRCh38
rs544747705 Chr.11: 109426636-109426636 on Build GRCh38
rs75343163 Chr.11: 109426696-109426696 on Build GRCh38
rs1029858665 Chr.11: 109426789-109426789 on Build GRCh38
rs951657826 Chr.11: 109426811-109426811 on Build GRCh38
rs1042215910 Chr.11: 109426665-109426665 on Build GRCh38
rs766606325 Chr.11: 109426842-109426842 on Build GRCh38
rs899823581 Chr.11: 109426754-109426754 on Build GRCh38
rs982968669 Chr.11: 109426421-109426421 on Build GRCh38
rs566912003 Chr.11: 109426418-109426418 on Build GRCh38
rs188749508 Chr.11: 109426600-109426600 on Build GRCh38
rs968531637 Chr.11: 109426673-109426673 on Build GRCh38
rs907156685 Chr.11: 109426561-109426561 on Build GRCh38
rs962500288 Chr.11: 109426870-109426870 on Build GRCh38
rs939251510 Chr.11: 109426601-109426601 on Build GRCh38
rs1039684592 Chr.11: 109426710-109426710 on Build GRCh38
rs908733713 Chr.11: 109426763-109426763 on Build GRCh38
rs568157430 Chr.11: 109426524-109426524 on Build GRCh38
rs765953708 Chr.11: 109426667-109426668 on Build GRCh38
rs181172613 Chr.11: 109426630-109426630 on Build GRCh38
rs373731789 Chr.11: 109426641-109426641 on Build GRCh38
rs759808008 Chr.11: 109426861-109426861 on Build GRCh38
rs773241773 Chr.11: 109426725-109426725 on Build GRCh38
rs531366835 Chr.11: 109426832-109426832 on Build GRCh38
rs1022346038 Chr.11: 109426640-109426640 on Build GRCh38
rs909614941 Chr.11: 109426484-109426486 on Build GRCh38
rs919719425 Chr.11: 109426839-109426839 on Build GRCh38
rs369901955 Chr.11: 109426628-109426628 on Build GRCh38
rs561253980 Chr.11: 109426818-109426818 on Build GRCh38
rs980456287 Chr.11: 109426556-109426556 on Build GRCh38

View Full Product Details