Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 109427502-109427995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf87
Entrez Gene ID
399947
Gene Name
chromosome 11 open reading frame 87
Gene Aliases
LOH11CR1A, NEURIM1
UniGene
Hs.172982
Gene Chromosome Location
Chr.11: 109422120-109433003 on Build GRCh38
Transcripts
NM_207645.3 XM_011542817.2 XM_011542818.2

SNP Detail

SNP ID Chromosome Location
rs1037061302 Chr.11: 109427729-109427729 on Build GRCh38
rs1019967127 Chr.11: 109427887-109427887 on Build GRCh38
rs988531375 Chr.11: 109427865-109427865 on Build GRCh38
rs190082174 Chr.11: 109427610-109427610 on Build GRCh38
rs550236885 Chr.11: 109427896-109427896 on Build GRCh38
rs959807526 Chr.11: 109427860-109427860 on Build GRCh38
rs921877672 Chr.11: 109427952-109427952 on Build GRCh38
rs370484376 Chr.11: 109427949-109427949 on Build GRCh38
rs964556278 Chr.11: 109427912-109427912 on Build GRCh38
rs545486943 Chr.11: 109427748-109427748 on Build GRCh38
rs754642136 Chr.11: 109427528-109427528 on Build GRCh38
rs1033467527 Chr.11: 109427700-109427700 on Build GRCh38
rs181697646 Chr.11: 109427765-109427765 on Build GRCh38
rs772543481 Chr.11: 109427659-109427659 on Build GRCh38
rs532915341 Chr.11: 109427730-109427730 on Build GRCh38
rs1013306075 Chr.11: 109427751-109427751 on Build GRCh38
rs377452428 Chr.11: 109427606-109427606 on Build GRCh38
rs931781044 Chr.11: 109427637-109427637 on Build GRCh38
rs527645152 Chr.11: 109427795-109427795 on Build GRCh38
rs553708312 Chr.11: 109427623-109427623 on Build GRCh38
rs371027175 Chr.11: 109427920-109427920 on Build GRCh38
rs987287561 Chr.11: 109427957-109427957 on Build GRCh38
rs540059736 Chr.11: 109427749-109427749 on Build GRCh38
rs1038997717 Chr.11: 109427633-109427633 on Build GRCh38
rs114873862 Chr.11: 109427837-109427837 on Build GRCh38
rs532372197 Chr.11: 109427702-109427702 on Build GRCh38
rs553413518 Chr.11: 109427966-109427966 on Build GRCh38
rs906108341 Chr.11: 109427774-109427774 on Build GRCh38
rs944136918 Chr.11: 109427964-109427964 on Build GRCh38
rs1051537885 Chr.11: 109427651-109427651 on Build GRCh38
rs187670338 Chr.11: 109427958-109427958 on Build GRCh38
rs565073003 Chr.11: 109427624-109427624 on Build GRCh38
rs966752036 Chr.11: 109427815-109427815 on Build GRCh38
rs912877193 Chr.11: 109427523-109427523 on Build GRCh38
rs947338312 Chr.11: 109427544-109427544 on Build GRCh38
rs1008247425 Chr.11: 109427662-109427662 on Build GRCh38
rs549360354 Chr.11: 109427802-109427802 on Build GRCh38
rs978708911 Chr.11: 109427950-109427950 on Build GRCh38
rs543390814 Chr.11: 109427611-109427611 on Build GRCh38
rs1001744806 Chr.11: 109427836-109427836 on Build GRCh38
rs779202868 Chr.11: 109427563-109427563 on Build GRCh38
rs532018294 Chr.11: 109427859-109427859 on Build GRCh38
rs974540495 Chr.11: 109427924-109427924 on Build GRCh38
rs897164732 Chr.11: 109427973-109427973 on Build GRCh38
rs770848850 Chr.11: 109427953-109427953 on Build GRCh38

View Full Product Details