Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 109427881-109428383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf87
Entrez Gene ID
399947
Gene Name
chromosome 11 open reading frame 87
Gene Aliases
LOH11CR1A, NEURIM1
UniGene
Hs.172982
Gene Chromosome Location
Chr.11: 109422120-109433003 on Build GRCh38
Transcripts
NM_207645.3 XM_011542817.2 XM_011542818.2

SNP Detail

SNP ID Chromosome Location
rs938192841 Chr.11: 109428020-109428020 on Build GRCh38
rs921877672 Chr.11: 109427952-109427952 on Build GRCh38
rs1009990326 Chr.11: 109428105-109428105 on Build GRCh38
rs964556278 Chr.11: 109427912-109427912 on Build GRCh38
rs1028596466 Chr.11: 109428134-109428134 on Build GRCh38
rs371027175 Chr.11: 109427920-109427920 on Build GRCh38
rs2298470 Chr.11: 109428302-109428302 on Build GRCh38
rs1001774517 Chr.11: 109428035-109428035 on Build GRCh38
rs535991319 Chr.11: 109428110-109428110 on Build GRCh38
rs931341947 Chr.11: 109428001-109428001 on Build GRCh38
rs987287561 Chr.11: 109427957-109427957 on Build GRCh38
rs748159088 Chr.11: 109428113-109428113 on Build GRCh38
rs1048903502 Chr.11: 109428295-109428295 on Build GRCh38
rs1055359232 Chr.11: 109428350-109428350 on Build GRCh38
rs937595672 Chr.11: 109428317-109428317 on Build GRCh38
rs146767274 Chr.11: 109428292-109428292 on Build GRCh38
rs191888207 Chr.11: 109428233-109428233 on Build GRCh38
rs2298471 Chr.11: 109428321-109428321 on Build GRCh38
rs940001868 Chr.11: 109427974-109427974 on Build GRCh38
rs986932157 Chr.11: 109428164-109428164 on Build GRCh38
rs954518009 Chr.11: 109428162-109428162 on Build GRCh38
rs1038638939 Chr.11: 109427992-109427992 on Build GRCh38
rs770169228 Chr.11: 109428182-109428182 on Build GRCh38
rs897164732 Chr.11: 109427973-109427973 on Build GRCh38
rs953323055 Chr.11: 109428139-109428139 on Build GRCh38
rs370484376 Chr.11: 109427949-109427949 on Build GRCh38
rs1048389084 Chr.11: 109428012-109428012 on Build GRCh38
rs558597119 Chr.11: 109428264-109428264 on Build GRCh38
rs911742139 Chr.11: 109428206-109428206 on Build GRCh38
rs554538286 Chr.11: 109428113-109428113 on Build GRCh38
rs898746365 Chr.11: 109428002-109428002 on Build GRCh38
rs1021112059 Chr.11: 109428069-109428069 on Build GRCh38
rs974592275 Chr.11: 109428133-109428133 on Build GRCh38
rs971917417 Chr.11: 109428234-109428234 on Build GRCh38
rs144771746 Chr.11: 109428114-109428114 on Build GRCh38
rs180965433 Chr.11: 109428286-109428286 on Build GRCh38
rs375626641 Chr.11: 109428324-109428324 on Build GRCh38
rs553413518 Chr.11: 109427966-109427966 on Build GRCh38
rs1030140976 Chr.11: 109428141-109428141 on Build GRCh38
rs905835657 Chr.11: 109428311-109428311 on Build GRCh38
rs776785014 Chr.11: 109428090-109428090 on Build GRCh38
rs944136918 Chr.11: 109427964-109427964 on Build GRCh38
rs1013358104 Chr.11: 109428043-109428043 on Build GRCh38
rs918707399 Chr.11: 109428258-109428258 on Build GRCh38
rs961439330 Chr.11: 109428210-109428210 on Build GRCh38
rs187670338 Chr.11: 109427958-109427958 on Build GRCh38
rs537908880 Chr.11: 109428353-109428356 on Build GRCh38
rs1055306221 Chr.11: 109428023-109428023 on Build GRCh38
rs978708911 Chr.11: 109427950-109427950 on Build GRCh38
rs978873542 Chr.11: 109428121-109428121 on Build GRCh38
rs974540495 Chr.11: 109427924-109427924 on Build GRCh38
rs115371703 Chr.11: 109428034-109428034 on Build GRCh38
rs895963747 Chr.11: 109428098-109428098 on Build GRCh38
rs770848850 Chr.11: 109427953-109427953 on Build GRCh38

View Full Product Details