Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 109428628-109429147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf87
Entrez Gene ID
399947
Gene Name
chromosome 11 open reading frame 87
Gene Aliases
LOH11CR1A, NEURIM1
UniGene
Hs.172982
Gene Chromosome Location
Chr.11: 109422120-109433003 on Build GRCh38
Transcripts
NM_207645.3 XM_011542817.2 XM_011542818.2

SNP Detail

SNP ID Chromosome Location
rs760670534 Chr.11: 109429029-109429029 on Build GRCh38
rs937289931 Chr.11: 109428833-109428833 on Build GRCh38
rs536660382 Chr.11: 109429119-109429119 on Build GRCh38
rs917543771 Chr.11: 109428848-109428848 on Build GRCh38
rs946301025 Chr.11: 109428849-109428849 on Build GRCh38
rs767550133 Chr.11: 109428933-109428933 on Build GRCh38
rs924094876 Chr.11: 109429046-109429047 on Build GRCh38
rs184940276 Chr.11: 109428732-109428732 on Build GRCh38
rs984181685 Chr.11: 109428754-109428754 on Build GRCh38
rs536328650 Chr.11: 109429019-109429019 on Build GRCh38
rs995779519 Chr.11: 109429115-109429115 on Build GRCh38
rs569602629 Chr.11: 109429094-109429094 on Build GRCh38
rs368358550 Chr.11: 109428988-109428988 on Build GRCh38
rs936436393 Chr.11: 109429072-109429072 on Build GRCh38
rs35162862 Chr.11: 109428906-109428907 on Build GRCh38
rs1018549236 Chr.11: 109429075-109429075 on Build GRCh38
rs532525097 Chr.11: 109428937-109428937 on Build GRCh38
rs78573448 Chr.11: 109428992-109428992 on Build GRCh38
rs900207622 Chr.11: 109429104-109429104 on Build GRCh38
rs746780141 Chr.11: 109428876-109428876 on Build GRCh38
rs920428574 Chr.11: 109428673-109428673 on Build GRCh38
rs548006485 Chr.11: 109429043-109429043 on Build GRCh38
rs1030191688 Chr.11: 109428662-109428662 on Build GRCh38
rs565975421 Chr.11: 109428993-109428993 on Build GRCh38
rs547584059 Chr.11: 109428965-109428965 on Build GRCh38
rs1050094200 Chr.11: 109428963-109428963 on Build GRCh38
rs571620205 Chr.11: 109428911-109428911 on Build GRCh38
rs991095767 Chr.11: 109428938-109428938 on Build GRCh38
rs750394486 Chr.11: 109428940-109428940 on Build GRCh38
rs888859548 Chr.11: 109429036-109429036 on Build GRCh38
rs904658230 Chr.11: 109428904-109428904 on Build GRCh38
rs372491245 Chr.11: 109429057-109429057 on Build GRCh38
rs1008558181 Chr.11: 109429042-109429042 on Build GRCh38
rs1042305119 Chr.11: 109428902-109428902 on Build GRCh38
rs919941953 Chr.11: 109428843-109428843 on Build GRCh38
rs927282753 Chr.11: 109428794-109428794 on Build GRCh38
rs1015711602 Chr.11: 109428768-109428768 on Build GRCh38
rs972333192 Chr.11: 109428653-109428653 on Build GRCh38
rs1053441162 Chr.11: 109429095-109429095 on Build GRCh38
rs949614687 Chr.11: 109428972-109428972 on Build GRCh38
rs1057145938 Chr.11: 109428837-109428837 on Build GRCh38
rs961766255 Chr.11: 109428824-109428824 on Build GRCh38
rs1042959585 Chr.11: 109428991-109428991 on Build GRCh38

View Full Product Details