Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128900242-128900747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
-
UniGene
Hs.689860
Gene Chromosome Location
Chr.11: 128899565-128909520 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

Gene Symbol
KCNJ5
Entrez Gene ID
3762
Gene Name
potassium voltage-gated channel subfamily J member 5
Gene Aliases
CIR, GIRK4, KATP1, KIR3.4, LQT13
UniGene
Hs.444595
Gene Chromosome Location
Chr.11: 128891418-128921163 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

SNP Detail

SNP ID Chromosome Location
rs544926307 Chr.11: 128900343-128900343 on Build GRCh38
rs184454632 Chr.11: 128900680-128900680 on Build GRCh38
rs995309879 Chr.11: 128900691-128900691 on Build GRCh38
rs774761334 Chr.11: 128900615-128900615 on Build GRCh38
rs908521169 Chr.11: 128900703-128900703 on Build GRCh38
rs745491371 Chr.11: 128900368-128900368 on Build GRCh38
rs779494660 Chr.11: 128900444-128900444 on Build GRCh38
rs1007582728 Chr.11: 128900622-128900622 on Build GRCh38
rs560151287 Chr.11: 128900520-128900520 on Build GRCh38
rs532239419 Chr.11: 128900524-128900524 on Build GRCh38
rs386758770 Chr.11: 128900578-128900579 on Build GRCh38
rs368671163 Chr.11: 128900341-128900341 on Build GRCh38
rs1017703012 Chr.11: 128900336-128900336 on Build GRCh38
rs963432001 Chr.11: 128900379-128900379 on Build GRCh38
rs771785065 Chr.11: 128900290-128900290 on Build GRCh38
rs976028459 Chr.11: 128900649-128900649 on Build GRCh38
rs2846698 Chr.11: 128900579-128900579 on Build GRCh38
rs965940879 Chr.11: 128900320-128900320 on Build GRCh38
rs1019715909 Chr.11: 128900313-128900313 on Build GRCh38
rs150171748 Chr.11: 128900549-128900549 on Build GRCh38
rs914054596 Chr.11: 128900575-128900575 on Build GRCh38
rs999955210 Chr.11: 128900577-128900577 on Build GRCh38
rs931330148 Chr.11: 128900354-128900354 on Build GRCh38
rs1026183682 Chr.11: 128900722-128900722 on Build GRCh38
rs552949720 Chr.11: 128900681-128900681 on Build GRCh38
rs372902460 Chr.11: 128900503-128900504 on Build GRCh38
rs1019851439 Chr.11: 128900610-128900610 on Build GRCh38
rs890453453 Chr.11: 128900299-128900299 on Build GRCh38
rs1009743788 Chr.11: 128900594-128900594 on Build GRCh38
rs984565170 Chr.11: 128900355-128900355 on Build GRCh38
rs904305035 Chr.11: 128900571-128900571 on Build GRCh38
rs879026389 Chr.11: 128900578-128900579 on Build GRCh38
rs3740835 Chr.11: 128900463-128900463 on Build GRCh38
rs2846699 Chr.11: 128900578-128900578 on Build GRCh38
rs965554798 Chr.11: 128900623-128900623 on Build GRCh38
rs558968086 Chr.11: 128900409-128900409 on Build GRCh38
rs76762583 Chr.11: 128900689-128900689 on Build GRCh38
rs921370656 Chr.11: 128900344-128900344 on Build GRCh38
rs528852096 Chr.11: 128900269-128900269 on Build GRCh38
rs572934413 Chr.11: 128900356-128900356 on Build GRCh38
rs192702506 Chr.11: 128900536-128900536 on Build GRCh38
rs184740558 Chr.11: 128900408-128900408 on Build GRCh38
rs569066591 Chr.11: 128900545-128900545 on Build GRCh38
rs925719713 Chr.11: 128900537-128900537 on Build GRCh38
rs192260762 Chr.11: 128900301-128900301 on Build GRCh38
rs917141606 Chr.11: 128900519-128900519 on Build GRCh38
rs994690695 Chr.11: 128900402-128900402 on Build GRCh38
rs566918368 Chr.11: 128900272-128900272 on Build GRCh38
rs575716981 Chr.11: 128900303-128900303 on Build GRCh38
rs188043755 Chr.11: 128900448-128900448 on Build GRCh38
rs1007531802 Chr.11: 128900334-128900334 on Build GRCh38

View Full Product Details