Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1335464-1335970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL1
Entrez Gene ID
1855
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
DRS2, DVL, DVL1L1, DVL1P1
UniGene
Hs.731450
Gene Chromosome Location
Chr.1: 1335278-1349142 on Build GRCh38
Transcripts
NM_004421.2 XM_005244731.3

SNP Detail

SNP ID Chromosome Location
rs541500383 Chr.1: 1335634-1335634 on Build GRCh38
rs569095502 Chr.1: 1335898-1335898 on Build GRCh38
rs186292265 Chr.1: 1335607-1335607 on Build GRCh38
rs1002967253 Chr.1: 1335628-1335628 on Build GRCh38
rs920935405 Chr.1: 1335709-1335709 on Build GRCh38
rs575030416 Chr.1: 1335674-1335674 on Build GRCh38
rs1049619436 Chr.1: 1335729-1335732 on Build GRCh38
rs991209782 Chr.1: 1335711-1335711 on Build GRCh38
rs947295787 Chr.1: 1335743-1335743 on Build GRCh38
rs148210175 Chr.1: 1335727-1335727 on Build GRCh38
rs903029045 Chr.1: 1335765-1335765 on Build GRCh38
rs541241468 Chr.1: 1335489-1335489 on Build GRCh38
rs147003803 Chr.1: 1335566-1335566 on Build GRCh38
rs886400446 Chr.1: 1335946-1335946 on Build GRCh38
rs1018602310 Chr.1: 1335635-1335635 on Build GRCh38
rs955810513 Chr.1: 1335846-1335846 on Build GRCh38
rs1035679069 Chr.1: 1335773-1335773 on Build GRCh38
rs1006776658 Chr.1: 1335617-1335617 on Build GRCh38
rs972311918 Chr.1: 1335810-1335810 on Build GRCh38
rs978500384 Chr.1: 1335692-1335692 on Build GRCh38
rs1016846952 Chr.1: 1335781-1335781 on Build GRCh38
rs912202590 Chr.1: 1335836-1335836 on Build GRCh38
rs753505839 Chr.1: 1335725-1335725 on Build GRCh38
rs994933201 Chr.1: 1335602-1335602 on Build GRCh38
rs912983306 Chr.1: 1335535-1335535 on Build GRCh38
rs967072449 Chr.1: 1335684-1335684 on Build GRCh38
rs761838615 Chr.1: 1335615-1335615 on Build GRCh38
rs529749618 Chr.1: 1335792-1335792 on Build GRCh38
rs978385581 Chr.1: 1335553-1335553 on Build GRCh38
rs901131015 Chr.1: 1335653-1335653 on Build GRCh38
rs540582999 Chr.1: 1335689-1335689 on Build GRCh38
rs190025538 Chr.1: 1335767-1335767 on Build GRCh38
rs763930844 Chr.1: 1335573-1335573 on Build GRCh38
rs933872961 Chr.1: 1335938-1335938 on Build GRCh38
rs532040477 Chr.1: 1335855-1335855 on Build GRCh38
rs1044285940 Chr.1: 1335747-1335747 on Build GRCh38
rs754280458 Chr.1: 1335762-1335763 on Build GRCh38
rs920916181 Chr.1: 1335491-1335491 on Build GRCh38
rs763973461 Chr.1: 1335923-1335923 on Build GRCh38
rs963984491 Chr.1: 1335690-1335690 on Build GRCh38
rs370650119 Chr.1: 1335785-1335785 on Build GRCh38
rs1057171859 Chr.1: 1335943-1335943 on Build GRCh38
rs954939316 Chr.1: 1335834-1335834 on Build GRCh38
rs749499869 Chr.1: 1335944-1335944 on Build GRCh38
rs975306112 Chr.1: 1335793-1335793 on Build GRCh38
rs901087451 Chr.1: 1335762-1335763 on Build GRCh38
rs143590035 Chr.1: 1335701-1335701 on Build GRCh38
rs1044618884 Chr.1: 1335913-1335913 on Build GRCh38
rs901872717 Chr.1: 1335877-1335877 on Build GRCh38
rs1011465911 Chr.1: 1335601-1335601 on Build GRCh38
rs950758725 Chr.1: 1335643-1335643 on Build GRCh38
rs997959139 Chr.1: 1335670-1335670 on Build GRCh38
rs566487590 Chr.1: 1335835-1335835 on Build GRCh38
rs1020051557 Chr.1: 1335764-1335764 on Build GRCh38
rs111452125 Chr.1: 1335759-1335759 on Build GRCh38
rs34391598 Chr.1: 1335657-1335657 on Build GRCh38
rs558442195 Chr.1: 1335544-1335544 on Build GRCh38
rs897896334 Chr.1: 1335600-1335600 on Build GRCh38
rs925622846 Chr.1: 1335555-1335556 on Build GRCh38
rs765828386 Chr.1: 1335720-1335720 on Build GRCh38
rs563066739 Chr.1: 1335721-1335721 on Build GRCh38
rs769819826 Chr.1: 1335860-1335860 on Build GRCh38
rs1053378008 Chr.1: 1335726-1335726 on Build GRCh38
rs930673784 Chr.1: 1335728-1335728 on Build GRCh38
rs61735961 Chr.1: 1335515-1335515 on Build GRCh38
rs950194505 Chr.1: 1335587-1335587 on Build GRCh38
rs769074137 Chr.1: 1335569-1335569 on Build GRCh38
rs1033171311 Chr.1: 1335783-1335783 on Build GRCh38
rs554217687 Chr.1: 1335697-1335697 on Build GRCh38
rs767621698 Chr.1: 1335681-1335681 on Build GRCh38
rs307373 Chr.1: 1335795-1335795 on Build GRCh38
rs1004947020 Chr.1: 1335778-1335778 on Build GRCh38
rs372736397 Chr.1: 1335854-1335854 on Build GRCh38
rs181921630 Chr.1: 1335503-1335503 on Build GRCh38
rs905825321 Chr.1: 1335616-1335616 on Build GRCh38
rs369800749 Chr.1: 1335585-1335585 on Build GRCh38
rs144058633 Chr.1: 1335713-1335713 on Build GRCh38
rs938550270 Chr.1: 1335926-1335926 on Build GRCh38
rs991547309 Chr.1: 1335789-1335789 on Build GRCh38
rs546627637 Chr.1: 1335680-1335680 on Build GRCh38
rs138223763 Chr.1: 1335593-1335593 on Build GRCh38
rs552433814 Chr.1: 1335880-1335880 on Build GRCh38
rs1041626746 Chr.1: 1335940-1335940 on Build GRCh38
rs767631787 Chr.1: 1335776-1335776 on Build GRCh38
rs1032641190 Chr.1: 1335693-1335693 on Build GRCh38
rs889242797 Chr.1: 1335746-1335746 on Build GRCh38
rs947275888 Chr.1: 1335876-1335876 on Build GRCh38
rs943495589 Chr.1: 1335857-1335857 on Build GRCh38
rs903145421 Chr.1: 1335942-1335942 on Build GRCh38
rs919389892 Chr.1: 1335829-1335831 on Build GRCh38
rs763113822 Chr.1: 1335763-1335763 on Build GRCh38
rs1020103249 Chr.1: 1335672-1335672 on Build GRCh38
rs538942382 Chr.1: 1335533-1335533 on Build GRCh38
rs747404588 Chr.1: 1335567-1335567 on Build GRCh38

View Full Product Details