Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1336061-1336558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL1
Entrez Gene ID
1855
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
DRS2, DVL, DVL1L1, DVL1P1
UniGene
Hs.731450
Gene Chromosome Location
Chr.1: 1335278-1349142 on Build GRCh38
Transcripts
NM_004421.2 XM_005244731.3 XM_005244732.3 XM_005244733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509650 c.1928delC p.P643fs*31 Chr.1: 1336302-1336302 on Build GRCh38
6225590 c.1850G>A p.R617Q Chr.1: 1336380-1336380 on Build GRCh38
6214851 c.2080A>G p.I694V Chr.1: 1336150-1336150 on Build GRCh38
400394 c.1785G>T p.P595P Chr.1: 1336370-1336370 on Build GRCh38
5544875 c.1734C>T p.S578S Chr.1: 1336496-1336496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769727331 Chr.1: 1336454-1336454 on Build GRCh38
rs374175996 Chr.1: 1336463-1336463 on Build GRCh38
rs767628325 Chr.1: 1336275-1336275 on Build GRCh38
rs375813716 Chr.1: 1336467-1336467 on Build GRCh38
rs774642394 Chr.1: 1336315-1336315 on Build GRCh38
rs774929779 Chr.1: 1336351-1336351 on Build GRCh38
rs763938693 Chr.1: 1336338-1336338 on Build GRCh38
rs376852453 Chr.1: 1336291-1336291 on Build GRCh38
rs745461448 Chr.1: 1336310-1336311 on Build GRCh38
rs769098890 Chr.1: 1336348-1336348 on Build GRCh38
rs377426716 Chr.1: 1336133-1336133 on Build GRCh38
rs758335347 Chr.1: 1336337-1336337 on Build GRCh38
rs977337066 Chr.1: 1336224-1336224 on Build GRCh38
rs953280751 Chr.1: 1336481-1336481 on Build GRCh38
rs760077387 Chr.1: 1336169-1336169 on Build GRCh38
rs768748298 Chr.1: 1336500-1336500 on Build GRCh38
rs766619185 Chr.1: 1336325-1336325 on Build GRCh38
rs757989455 Chr.1: 1336474-1336474 on Build GRCh38
rs370626248 Chr.1: 1336505-1336505 on Build GRCh38
rs760119618 Chr.1: 1336123-1336123 on Build GRCh38
rs760791785 Chr.1: 1336319-1336319 on Build GRCh38
rs775980552 Chr.1: 1336255-1336255 on Build GRCh38
rs777173993 Chr.1: 1336168-1336168 on Build GRCh38
rs372681220 Chr.1: 1336453-1336453 on Build GRCh38
rs905441386 Chr.1: 1336433-1336433 on Build GRCh38
rs769030905 Chr.1: 1336314-1336314 on Build GRCh38
rs377189378 Chr.1: 1336419-1336419 on Build GRCh38
rs756990609 Chr.1: 1336261-1336262 on Build GRCh38
rs774183099 Chr.1: 1336121-1336121 on Build GRCh38
rs749297714 Chr.1: 1336492-1336492 on Build GRCh38
rs755030520 Chr.1: 1336217-1336217 on Build GRCh38
rs779009859 Chr.1: 1336222-1336222 on Build GRCh38
rs756416367 Chr.1: 1336486-1336486 on Build GRCh38
rs566523601 Chr.1: 1336146-1336146 on Build GRCh38
rs182333546 Chr.1: 1336482-1336482 on Build GRCh38
rs765583811 Chr.1: 1336171-1336171 on Build GRCh38
rs568283927 Chr.1: 1336366-1336366 on Build GRCh38
rs763319960 Chr.1: 1336173-1336173 on Build GRCh38
rs769935706 Chr.1: 1336411-1336411 on Build GRCh38
rs768600525 Chr.1: 1336098-1336098 on Build GRCh38
rs763742739 Chr.1: 1336532-1336532 on Build GRCh38
rs759267143 Chr.1: 1336523-1336523 on Build GRCh38
rs866234681 Chr.1: 1336452-1336452 on Build GRCh38
rs775705276 Chr.1: 1336456-1336456 on Build GRCh38
rs753552772 Chr.1: 1336426-1336426 on Build GRCh38
rs753145330 Chr.1: 1336434-1336434 on Build GRCh38
rs142013934 Chr.1: 1336410-1336410 on Build GRCh38
rs779576393 Chr.1: 1336343-1336343 on Build GRCh38
rs774130151 Chr.1: 1336413-1336413 on Build GRCh38
rs750475116 Chr.1: 1336281-1336281 on Build GRCh38
rs61757368 Chr.1: 1336472-1336472 on Build GRCh38
rs142476987 Chr.1: 1336375-1336375 on Build GRCh38
rs925302561 Chr.1: 1336218-1336218 on Build GRCh38
rs750000072 Chr.1: 1336529-1336529 on Build GRCh38
rs748424414 Chr.1: 1336353-1336353 on Build GRCh38
rs147591285 Chr.1: 1336406-1336406 on Build GRCh38
rs556292877 Chr.1: 1336376-1336376 on Build GRCh38
rs753792159 Chr.1: 1336209-1336209 on Build GRCh38
rs751779370 Chr.1: 1336262-1336262 on Build GRCh38
rs374846579 Chr.1: 1336330-1336330 on Build GRCh38
rs774094398 Chr.1: 1336099-1336099 on Build GRCh38
rs761979060 Chr.1: 1336269-1336269 on Build GRCh38
rs753401275 Chr.1: 1336471-1336471 on Build GRCh38
rs372041252 Chr.1: 1336373-1336373 on Build GRCh38
rs764309779 Chr.1: 1336196-1336196 on Build GRCh38
rs374440563 Chr.1: 1336225-1336225 on Build GRCh38
rs771937584 Chr.1: 1336461-1336461 on Build GRCh38
rs570904822 Chr.1: 1336374-1336374 on Build GRCh38
rs907783331 Chr.1: 1336090-1336090 on Build GRCh38
rs779371053 Chr.1: 1336537-1336537 on Build GRCh38
rs750042711 Chr.1: 1336379-1336379 on Build GRCh38
rs776909760 Chr.1: 1336371-1336371 on Build GRCh38
rs368265757 Chr.1: 1336403-1336403 on Build GRCh38
rs781647592 Chr.1: 1336483-1336483 on Build GRCh38
rs1030468130 Chr.1: 1336301-1336302 on Build GRCh38
rs781175326 Chr.1: 1336201-1336201 on Build GRCh38
rs751439375 Chr.1: 1336146-1336148 on Build GRCh38
rs556813803 Chr.1: 1336079-1336079 on Build GRCh38
rs745542260 Chr.1: 1336341-1336341 on Build GRCh38
rs370205492 Chr.1: 1336370-1336370 on Build GRCh38
rs765639162 Chr.1: 1336132-1336132 on Build GRCh38
rs778386571 Chr.1: 1336480-1336480 on Build GRCh38
rs545740677 Chr.1: 1336524-1336524 on Build GRCh38
rs1038087714 Chr.1: 1336108-1336108 on Build GRCh38
rs867309666 Chr.1: 1336180-1336180 on Build GRCh38
rs776073474 Chr.1: 1336239-1336239 on Build GRCh38
rs531416128 Chr.1: 1336226-1336226 on Build GRCh38
rs759036207 Chr.1: 1336436-1336436 on Build GRCh38
rs772538439 Chr.1: 1336397-1336397 on Build GRCh38
rs539920742 Chr.1: 1336160-1336160 on Build GRCh38
rs748686111 Chr.1: 1336381-1336381 on Build GRCh38
rs757905037 Chr.1: 1336534-1336534 on Build GRCh38
rs780834482 Chr.1: 1336302-1336302 on Build GRCh38
rs956729494 Chr.1: 1336308-1336308 on Build GRCh38
rs779242416 Chr.1: 1336387-1336387 on Build GRCh38
rs572292192 Chr.1: 1336451-1336451 on Build GRCh38
rs372749605 Chr.1: 1336416-1336416 on Build GRCh38
rs767102382 Chr.1: 1336120-1336120 on Build GRCh38
rs747313105 Chr.1: 1336445-1336445 on Build GRCh38
rs768545790 Chr.1: 1336142-1336142 on Build GRCh38
rs778441474 Chr.1: 1336438-1336438 on Build GRCh38
rs769324780 Chr.1: 1336304-1336304 on Build GRCh38
rs376204834 Chr.1: 1336200-1336200 on Build GRCh38
rs746689239 Chr.1: 1336228-1336228 on Build GRCh38
rs760196398 Chr.1: 1336163-1336163 on Build GRCh38
rs997739969 Chr.1: 1336394-1336394 on Build GRCh38
rs749795230 Chr.1: 1336347-1336347 on Build GRCh38
rs113005319 Chr.1: 1336473-1336473 on Build GRCh38
rs370097001 Chr.1: 1336307-1336307 on Build GRCh38
rs755595194 Chr.1: 1336342-1336342 on Build GRCh38
rs915847905 Chr.1: 1336230-1336230 on Build GRCh38
rs761603875 Chr.1: 1336106-1336106 on Build GRCh38
rs772591531 Chr.1: 1336354-1336354 on Build GRCh38
rs146336697 Chr.1: 1336151-1336151 on Build GRCh38
rs988595463 Chr.1: 1336522-1336522 on Build GRCh38
rs747052391 Chr.1: 1336138-1336138 on Build GRCh38
rs139073062 Chr.1: 1336139-1336139 on Build GRCh38
rs770226805 Chr.1: 1336243-1336243 on Build GRCh38
rs780836585 Chr.1: 1336289-1336290 on Build GRCh38
rs531643983 Chr.1: 1336316-1336316 on Build GRCh38
rs940589568 Chr.1: 1336215-1336215 on Build GRCh38
rs1057038766 Chr.1: 1336089-1336089 on Build GRCh38
rs780781683 Chr.1: 1336339-1336339 on Build GRCh38
rs758190734 Chr.1: 1336292-1336292 on Build GRCh38
rs1021697382 Chr.1: 1336167-1336167 on Build GRCh38
rs935943551 Chr.1: 1336088-1336088 on Build GRCh38
rs377142799 Chr.1: 1336398-1336398 on Build GRCh38
rs778958560 Chr.1: 1336289-1336289 on Build GRCh38
rs760024382 Chr.1: 1336240-1336240 on Build GRCh38
rs746990390 Chr.1: 1336518-1336518 on Build GRCh38
rs140108185 Chr.1: 1336380-1336380 on Build GRCh38
rs938582015 Chr.1: 1336084-1336084 on Build GRCh38
rs374246352 Chr.1: 1336491-1336491 on Build GRCh38
rs748742257 Chr.1: 1336392-1336392 on Build GRCh38
rs776551474 Chr.1: 1336520-1336520 on Build GRCh38
rs759492048 Chr.1: 1336409-1336409 on Build GRCh38
rs766847400 Chr.1: 1336126-1336126 on Build GRCh38
rs34935417 Chr.1: 1336256-1336256 on Build GRCh38
rs770444542 Chr.1: 1336238-1336238 on Build GRCh38
rs899274517 Chr.1: 1336111-1336111 on Build GRCh38
rs562357128 Chr.1: 1336525-1336525 on Build GRCh38
rs771450757 Chr.1: 1336446-1336446 on Build GRCh38
rs773112898 Chr.1: 1336514-1336514 on Build GRCh38
rs202097902 Chr.1: 1336468-1336468 on Build GRCh38
rs770627955 Chr.1: 1336519-1336519 on Build GRCh38
rs143613619 Chr.1: 1336428-1336428 on Build GRCh38
rs752707849 Chr.1: 1336290-1336290 on Build GRCh38
rs1031248155 Chr.1: 1336150-1336150 on Build GRCh38
rs773602516 Chr.1: 1336318-1336318 on Build GRCh38
rs1061334 Chr.1: 1336391-1336391 on Build GRCh38
rs983325033 Chr.1: 1336448-1336448 on Build GRCh38
rs529716214 Chr.1: 1336157-1336157 on Build GRCh38
rs996637659 Chr.1: 1336131-1336131 on Build GRCh38
rs368143616 Chr.1: 1336367-1336367 on Build GRCh38
rs562753230 Chr.1: 1336293-1336293 on Build GRCh38
rs368894487 Chr.1: 1336449-1336449 on Build GRCh38
rs754549970 Chr.1: 1336385-1336385 on Build GRCh38
rs1035605245 Chr.1: 1336521-1336521 on Build GRCh38
rs770548436 Chr.1: 1336301-1336301 on Build GRCh38
rs551930213 Chr.1: 1336282-1336282 on Build GRCh38
rs946996007 Chr.1: 1336412-1336412 on Build GRCh38
rs146143161 Chr.1: 1336404-1336404 on Build GRCh38
rs1032029104 Chr.1: 1336296-1336296 on Build GRCh38
rs887456356 Chr.1: 1336278-1336278 on Build GRCh38
rs770370467 Chr.1: 1336166-1336166 on Build GRCh38
rs892749539 Chr.1: 1336283-1336283 on Build GRCh38
rs371155444 Chr.1: 1336223-1336223 on Build GRCh38
rs1047404649 Chr.1: 1336266-1336266 on Build GRCh38
rs1061335 Chr.1: 1336365-1336365 on Build GRCh38
rs1034189465 Chr.1: 1336189-1336189 on Build GRCh38
rs201423710 Chr.1: 1336526-1336526 on Build GRCh38
rs186463928 Chr.1: 1336134-1336134 on Build GRCh38
rs755656513 Chr.1: 1336535-1336535 on Build GRCh38
rs1061336 Chr.1: 1336313-1336313 on Build GRCh38
rs926897012 Chr.1: 1336530-1336530 on Build GRCh38
rs373403505 Chr.1: 1336127-1336127 on Build GRCh38
rs145478154 Chr.1: 1336418-1336418 on Build GRCh38

View Full Product Details